Skip to Content
Merck
All Photos(1)

Key Documents

33577

Supelco

Microcystin-RR solution

10 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H75N13O12
CAS Number:
Molecular Weight:
1038.20
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@@H](Cc1ccccc1)[C@@H](C)\C=C(C)\C=C\[C@@H]2NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](C)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](C)NC(=O)C(=C)N(C)C(=O)CC[C@@H](NC(=O)[C@H]2C)C(O)=O)C(O)=O

InChI

1S/C49H75N13O12/c1-26(24-27(2)37(74-8)25-32-14-10-9-11-15-32)18-19-33-28(3)40(64)60-36(46(70)71)20-21-38(63)62(7)31(6)43(67)56-30(5)42(66)59-35(17-13-23-55-49(52)53)45(69)61-39(47(72)73)29(4)41(65)58-34(44(68)57-33)16-12-22-54-48(50)51/h9-11,14-15,18-19,24,27-30,33-37,39H,6,12-13,16-17,20-23,25H2,1-5,7-8H3,(H,56,67)(H,57,68)(H,58,65)(H,59,66)(H,60,64)(H,61,69)(H,70,71)(H,72,73)(H4,50,51,54)(H4,52,53,55)/b19-18+,26-24+/t27-,28-,29-,30+,33-,34-,35-,36+,37-,39+/m0/s1

InChI key

JIGDOBKZMULDHS-UUHBQKJESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Microcystin-RR is a microcystin generally produced by cyanobacteria.

Application

Microcystin-RR may be used as an analytical reference standard for the quantification of the analyte in water samples using high-performance liquid chromatography with diode array detection (HPLC-DAD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heng-feng Miao et al.
Huan jing ke xue= Huanjing kexue, 31(5), 1239-1245 (2010-07-14)
Degradation of algal toxin Microcystin-RR (MC-RR) by ozonation processes was investigated. The degradation rate of MC-RR reached 83.0% at ozone/MC-RR dosage of 6, and the degradation efficiency was decreased with increase of pH or NOM contents. Ozonation byproducts of MC-RR
Chunyong Zhang et al.
Journal of hazardous materials, 172(2-3), 847-853 (2009-08-21)
Microcystins (MCs), produced by blue-green algae, are one of the most common naturally occurring toxins found in natural environment. The presence of MCs in drinking water sources poses a great threat to people's health. In this study, the degradation behavior
Gaodao Liang et al.
TheScientificWorldJournal, 11, 20-26 (2011-01-11)
Microcystins (MCs) are well known worldwide as hepatotoxins produced by cyanobacteria, but little is known about the physicochemical properties of these compounds. The dependence of the n-octanol/water distribution ratio (DOW) of MC-RR and -LR to pH was measured by high-performance
Mulan Zhang et al.
Journal of environmental sciences (China), 22(2), 168-175 (2010-04-20)
A strain, USTB-05, isolated from Lake Dianchi, China, degraded the cyanobacterial toxin microcystin-RR (MC-RR) at the rate of 16.7 mg/L per day. Analysis of 16S rDNA sequence showed that the strain was Sphingopyxis sp. Enzymatic degradation pathways for MC-RR by
Peng Chen et al.
Environmental science & technology, 46(4), 2345-2351 (2012-01-19)
Microcystin-RR (MC-RR) is one of the most common cyanotoxin microcystins in fresh water and is of great concern due to its potential hepatotoxicity. In the present study, Bi(2)WO(6) was synthesized with a hydrothermal method by varying the pH of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service