Skip to Content
Merck
All Photos(2)

Key Documents

33388

Supelco

Phorate

PESTANAL®, analytical standard

Synonym(s):

O,O-Diethyl S-(ethylthiomethyl) phosphorodithioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17O2PS3
CAS Number:
Molecular Weight:
260.38
Beilstein:
1708517
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SCSCC

InChI

1S/C7H17O2PS3/c1-4-8-10(11,9-5-2)13-7-12-6-3/h4-7H2,1-3H3

InChI key

BULVZWIRKLYCBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phorate is an organophosphate insecticide used to control sap-feeding insects including various beetles, mites, grubs, and worms in agricultural crop cultivation.

Application

Phorate may be used as a reference standard for the determination of the analyte in peanut oil, soybean oil and sesame oil using gas chromatography coupled with flame photometric detector (GC-FPD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Changkun Li et al.
Frontiers in microbiology, 9, 2048-2048 (2018-09-21)
This work performed a large scale assessment for organophosphorus pesticides (OPPs) degradation activity of 121 Lactobacillus (L.) plantarum strains. Six L. plantarum strains (P9, IMAU80110, IMAU40100, IMAU10585, IMAU10209, and IMAU80070) were found to possess high capacity of degrading three commonly
Determination of organophosphorus pesticides in soybean oil, peanut oil and sesame oil by low-temperature extraction and GC-FPD.
Li L, et al.
Chromatographia, 66(7-8), 625-629 (2007)
Yuanyuan Zhang et al.
Ultrasonics sonochemistry, 19(1), 43-48 (2011-06-15)
The degradation of phorate in apple juice by sonication was investigated in the present study. Results showed that sonication was effective in eliminating phorate in apple juice, and the ultrasonic power and sonication time significantly influenced the degradation of phorate
Amal Chandra Das et al.
Chemosphere, 58(5), 579-584 (2004-12-29)
An experiment has been conducted under laboratory conditions to investigate the effect of phorate (an organophosphate insecticide) and carbofuran (a carbamate insecticide) at their recommended field rates (1.5 and 1.0 kga.i.ha-1, respectively) on the growth and multiplication of microorganisms as
G Mohanty et al.
Ecotoxicology (London, England), 20(1), 283-292 (2010-12-15)
Labeo rohita (rohu) fingerlings were exposed to different concentrations (0.001, 0.002 and 0.01 ppm) of phorate, an organophosphate pesticide; samplings were done at 24, 48, 72 and 96 h. The study was carried out to evaluate tissue specific genotoxic effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service