Skip to Content
Merck
All Photos(2)

Key Documents

32097

Supelco

Etoricoxib

VETRANAL®, analytical standard

Synonym(s):

5-Chloro-3-[4-(methylsulfonyl)phenyl]-2-(2-methyl-5-pyridinyl)pyridine, 5-Chloro-6′-methyl-3-[4-(methylsulfonyl)phenyl]-2,3′-bipyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15ClN2O2S
CAS Number:
Molecular Weight:
358.84
Beilstein:
8073797
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CC1=NC=C(C2=C(C3=CC=C(S(C)(=O)=O)C=C3)C=C(Cl)C=N2)C=C1

InChI

1S/C18H15ClN2O2S/c1-12-3-4-14(10-20-12)18-17(9-15(19)11-21-18)13-5-7-16(8-6-13)24(2,22)23/h3-11H,1-2H3

InChI key

MNJVRJDLRVPLFE-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

General description

Etoricoxib is a non-steroidal cyclooxygenase-2 (COX-2) selective anti-inflammatory drug mostly used to treat patients suffering from osteoarthritis and rheumatoid arthritis.

Application

Etoricoxib may have been used as reference standard in liquid chromatography-tandem mass spectrometry method with atmospheric pressure chemical ionization (LC-APCI/MS/MS) method for determination of etoricoxib from human plasma.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Deborah J Cochrane et al.
Drugs, 62(18), 2637-2651 (2002-12-06)
Etoricoxib is a cyclo-oxygenase (COX)-2-selective NSAID with a higher COX-1 to COX-2 selectivity ratio than the other COX-2-selective NSAIDs rofecoxib, valdecoxib or celecoxib. In patients with rheumatoid arthritis, improvements in tender and swollen joint counts and patient and investigator global
Klaus Schaffler et al.
British journal of clinical pharmacology, 75(2), 404-414 (2012-07-11)
Laser (radiant-heat) evoked potentials (LEPs) from vertex-EEG peak-to-peak (PtP) amplitude were used to determine acute antinociceptive/antihyperalgesic efficacy of ABT-102, a novel TRPV1 antagonist efficacious in preclinical pain models, compared with active controls and placebo in normal and UV(B)-inflamed skin. This
Rachel Clarke et al.
The Cochrane database of systematic reviews, 4(4), CD004309-CD004309 (2012-04-20)
Etoricoxib is a selective cyclo-oxygenase-2 (COX-2) inhibitor licensed for the relief of chronic pain in osteoarthritis and rheumatoid arthritis, and acute pain in some jurisdictions. This class of drugs is believed to be associated with fewer upper gastrointestinal adverse effects
Paola Patrignani et al.
Expert opinion on pharmacotherapy, 4(2), 265-284 (2003-02-04)
The development of COX2 inhibitors with improved biochemical selectivity (such as etoricoxib and valdecoxib) over that of commercially available coxibs has been driven by the potential advantage of safety using higher coxib doses for increased efficacy. Etoricoxib has been approved
Rachel Clarke et al.
The Cochrane database of systematic reviews, (2)(2), CD004309-CD004309 (2009-04-17)
Etoricoxib is a selective cyclo-oxygenase-2 (COX-2) inhibitor prescribed for the relief of chronic pain in osteoarthritis and rheumatoid arthritis, and acute pain. The drug is believed to be associated with fewer upper gastrointestinal adverse effects than conventional non-steroidal anti-inflammatory drugs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service