Skip to Content
Merck
All Photos(1)

Documents

95594

Sigma-Aldrich

4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde

mixture of isomers, ≥97.0% (GC)

Synonym(s):

4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde, 4-(4-Hydroxy-4-methylpentyl)-3-cyclohexenecarboxaldehyde, 4-(4-Methyl-4-hydroxyamyl)cyclohex-3-ene carboxaldehyde, Landolal, Liral

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O2
CAS Number:
Molecular Weight:
210.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

density

0.995 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[H]C(=O)C1CCC(CCCC(C)(C)O)=CC1

InChI

1S/C13H22O2/c1-13(2,15)9-3-4-11-5-7-12(10-14)8-6-11/h5,10,12,15H,3-4,6-9H2,1-2H3

InChI key

ORMHZBNNECIKOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

275.2 °F

Flash Point(C)

135.1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is hydroxyisohexyl 3-cyclohexene carboxaldehyde sensitization declining in central Europe?
Axel Schnuch et al.
Contact dermatitis, 67(1), 47-49 (2012-06-12)
W Uter et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 21(3), 374-379 (2007-02-21)
Assessment of the value of patch testing patients' own perfumes, eau de toilette, deodorants and shaving lotions with regard to diagnosing contact allergy to fragrances, and an analysis of the spectrum of concurrent patch test reactions to single fragrance allergens.
Maria V Heisterberg et al.
Contact dermatitis, 63(5), 270-276 (2010-10-16)
Fragrance mix II (FM II) is a relatively new screening marker for fragrance contact allergy. It was introduced in the patch test baseline series in Denmark in 2005 and contains six different fragrance chemicals commonly present in cosmetic products and
Maria V Heisterberg et al.
Contact dermatitis, 65(5), 266-275 (2011-09-29)
Fragrance ingredients are a frequent cause of allergic contact dermatitis. The EU Cosmetics Directive states that 26 specific fragrance ingredients, known to cause allergic contact dermatitis, must be declared on the ingredient lists of cosmetic products. To investigate frequencies of
Rodrigo Carvalho et al.
Cutaneous and ocular toxicology, 30(3), 249-250 (2011-02-25)
Hydroxyisohexyl 3-cyclohexene carboxaldehyde (HICC)-Lyral® is a widely used synthetic fragrance found in personal care and household products. It is an aldehyde, lipophilic enough to penetrate the skin and is a frequent cause of contact sensitization. Describe the frequency of contact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service