Skip to Content
Merck
All Photos(1)

Key Documents

32061

Supelco

Trifluralin

PESTANAL®, analytical standard

Synonym(s):

2,6-Dinitro-N,N-dipropyl-4-trifluoromethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16F3N3O4
CAS Number:
Molecular Weight:
335.28
Beilstein:
1893555
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCN(CCC)c1c(cc(cc1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

InChI

1S/C13H16F3N3O4/c1-3-5-17(6-4-2)12-10(18(20)21)7-9(13(14,15)16)8-11(12)19(22)23/h7-8H,3-6H2,1-2H3

InChI key

ZSDSQXJSNMTJDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trifluralin is a dinitroaniline herbicide used mostly as antiparasitic agent. It is used in controlling weeds and is characterized as microtubule-depolymerizing chemical.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Trifluralin may have been used as standard in mass spectrometry employing negative chemical
ionization (GC-MS-NCI) technique for qualitative and quantitative analysis of trifluralin from soil extract samples.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quantitative confirmation of ethalfluralin and trifluralin in soil extracts by negative chemical ionization mass spectrometry.
Moyer, James R., and James L. Elder.
Journal of Agricultural and Food Chemistry, 32.4, 866-868 (1984)
Inhibition of leishmanias but not host macrophages by the antitubulin herbicide trifluralin.
Chan, Marion Man-Ying, and Dunne Fong.
Science, 249.4971, 924-924 (1990)
Mechanism of micronuclei formation in polyploidizated cells of Allium cepa exposed to trifluralin herbicide.
Fernandes, Thais CC, Dania Elisa C. Mazzeo, and Maria A. Marin-Morales.
Pesticide Biochemistry and Physiology, 88.3, 252-259 (2007)
V K Shormanov et al.
Sudebno-meditsinskaia ekspertiza, 53(5), 31-33 (2011-01-27)
Results of extraction of trifluralin from an aqueous acetonitrile solution using various organic solvents are presented. The degree of extraction was shown to depend on the nature of extractant and the water/acetonitrile ratio. An optimal electrolyte and the degree of
Perihan Binnur Kurt-Karakus et al.
Environmental toxicology and chemistry, 30(7), 1539-1548 (2011-04-08)
Concentrations of current-use pesticides (CUPs) in water, zooplankton, precipitation, and air samples as well as stereoisomer fractions (SF; herbicidally active/total stereoisomers) of metolachlor were determined in water samples collected from 10 remote inland lakes in Ontario, Canada, between 2003 and

Articles

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service