Skip to Content
Merck
All Photos(1)

Key Documents

31251

Supelco

Pretilachlor

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-2′,6′-diethyl-N-(2-propoxyethyl)acetanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26ClNO2
CAS Number:
Molecular Weight:
311.85
Beilstein:
2754162
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCOCCN(C(=O)CCl)c1c(CC)cccc1CC

InChI

1S/C17H26ClNO2/c1-4-11-21-12-10-19(16(20)13-18)17-14(5-2)8-7-9-15(17)6-3/h7-9H,4-6,10-13H2,1-3H3

InChI key

YLPGTOIOYRQOHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen-Jiang Liu et al.
Ecotoxicology and environmental safety, 74(6), 1595-1599 (2011-05-03)
An indirect competitive enzyme-linked immunosorbent assay (ic-ELISA) has been developed for detection of pretilachlor in water and soil. An immunogen was prepared from haptens of pretilachlor conjugated to bovine serum albumin(BSA). The specific polyclonal antibodies were obtained by immunizing New
Irani Mukherjee et al.
Bulletin of environmental contamination and toxicology, 100(6), 856-861 (2018-04-20)
This study was conducted at ambient (398 ± 10 µmol mol-1), elevated (450 ± 10 µmol mol-1) and elevated (550 ± 10 µmol mol-1) atmospheric CO2 under three moisture regime and also three level of temperature (4, 25, and 40°C) to assess the degradation of pretilachlor and butachlor. Under dry condition at
Luciano Scarponi et al.
Pest management science, 61(4), 371-376 (2005-03-08)
The persistence of pretilachlor applied to rice singly or in combination with the safener fenclorim was investigated in connection with starch, glucose and protein formation. The addition of fenclorim to pretilachlor did not reduce the accumulation of the latter, but
Najmeh Razzaghi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 120, 32-40 (2018-07-11)
This study was undertaken to analyze 29 pesticides residues in 37 commercially olive oil collected samples from Iran's markets using Quick, Easy, Cheap, Effective, Rugged, and Safe (QuEChERS) approach along with acetonitrile for the extraction, surface adsorbents for clean-up procedure
Pervinder Kaur et al.
Environmental technology, 38(19), 2410-2415 (2016-11-22)
The effect of long-term application of pretilachlor to paddy in rice-wheat cropping system was investigated from 1997 to 2015. Additionally, in 2013, field experiment was also conducted where pretilachlor was applied to paddy field having no background of its application.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service