Skip to Content
Merck
All Photos(1)

Key Documents

30122

Sigma-Aldrich

L-Cysteine hydrochloride monohydrate

tested according to Ph. Eur.

Synonym(s):

Cysteini hydrochloridum monohydricum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
Beilstein:
5158059
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

Assay

98.5-101.0%

form

solid

mp

176 °C

solubility

H2O: soluble 100 g/L at 25 °C

application(s)

pharmaceutical (small molecule)

SMILES string

O.Cl.N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m0../s1

InChI key

QIJRTFXNRTXDIP-JIZZDEOASA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Cysteine hydrochloride monohydrate was used in the synthesis of polycarbophil-cysteine conjugate.

Biochem/physiol Actions

Cysteine is a non-essential, thiol-containing amino acid that has protective effects in normal tissues. It is converted to glutathione that decreases the effects of chemotherapeutic and radiation agents. This might restrict the effect of therapeutic agents used for cancer treatment.
NMDA glutamatergic receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Marschütz et al.
Pharmaceutical research, 17(12), 1468-1474 (2001-04-17)
To develop an oral controlled release system for insulin. The polymer-inhibitor conjugates carboxymethylcellulose (CMC)-Bowman-Birk inhibitor and CMC-elastatinal were homogenized with polycarbophil-cysteine conjugate, insulin, and mannitol, compressed to 2 mg microtablets and enteric coated with a polymethacrylate. The protective effect of
J C Roberts
Amino acids, 8(2), 113-124 (1995-06-01)
The thiol-containing amino acid, cysteine, and its analogs are useful for a variety of protective applications, including protecting normal tissues against the unwanted side effects of cancer chemotherapeutic agents and radiation treatment. The protection can result from both direct action
Matthew Oser et al.
Journal of cell science, 123(Pt 21), 3662-3673 (2010-10-26)
Invadopodia are matrix-degrading membrane protrusions in invasive carcinoma cells enriched in proteins that regulate actin polymerization. The on-off regulatory switch that initiates actin polymerization in invadopodia requires phosphorylation of tyrosine residues 421, 466, and 482 on cortactin. However, it is
Qinjian Zhao et al.
Human vaccines & immunotherapeutics, 10(3), 734-739 (2013-12-05)
Cryo-transmission electron microscopy (cryoTEM) is a powerful characterization method for assessing the structural properties of biopharmaceutical nanoparticles, including Virus Like Particle-based vaccines. We demonstrate the method using the Human Papilloma Virus (HPV) VLPs in GARDASIL®. CryoTEM, coupled to automated data
Edward T Chouchani et al.
Nature medicine, 19(6), 753-759 (2013-05-28)
Oxidative damage from elevated production of reactive oxygen species (ROS) contributes to ischemia-reperfusion injury in myocardial infarction and stroke. The mechanism by which the increase in ROS occurs is not known, and it is unclear how this increase can be

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service