Skip to Content
Merck
L-Cysteine hydrochloride monohydrate tested according to Ph. Eur.

L-Cysteine hydrochloride monohydrate

L-Cysteine hydrochloride monohydrate tested according to Ph. Eur.
Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
API Family
Reference Material Type
Special Grade
Technique