Skip to Content
Merck
All Photos(1)

Key Documents

27483

Supelco

(−)-β-Citronellol

analytical standard

Synonym(s):

β-Rhodinol, (S)-(−)-β-Citronellol, (S)-3,7-Dimethyl-6-octen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
1721505
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D −5.3±0.2°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.456

bp

221-224 °C

density

0.854 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C[C@H](CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m0/s1

InChI key

QMVPMAAFGQKVCJ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Citronellol is a monoterpene alcohol found in essential oils of Cymbopogon winterianus Jowitt. S-(−)-isomer that is less common. It is prevalent in geranium and citronella oils.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Chiral building block

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K. Mori et al.
Liebigs Ann. Chem., 717-717 (1988)
Study of anticonvulsant effect of citronellol, a monoterpene alcohol, in rodents.
de Sousa DP, et al.
Neuroscience Letters, 401(3), 231-235 (2006)
Renan G Brito et al.
Basic & clinical pharmacology & toxicology, 112(4), 215-221 (2012-10-06)
Citronellol (CT) is a monoterpenoid alcohol present in the essential oil of many medicinal plants, such as Cymbopogon citratus. We evaluated the antinociceptive effects of CT on orofacial nociception in mice and investigated the central pathway involved in the effect.
M Spiller et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 120(7), 1461-1471 (2010-01-20)
The scent of flowers is a very important trait in ornamental roses in terms of both quantity and quality. In cut roses, scented varieties are a rare exception. Although metabolic profiling has identified more than 500 scent volatiles from rose
Joana F A Bastos et al.
Basic & clinical pharmacology & toxicology, 106(4), 331-337 (2009-12-17)
Citronellol is an essential oil constituent from the medicinal plants Cymbopogon citratus, Cymbopogon winterianus and Lippia alba which are thought to possess antihypertensive properties. Citronellol-induced cardiovascular effects were evaluated in this study. In rats, citronellol (1-20 mg/kg, i.v.) induced hypotension

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service