Skip to Content
Merck
All Photos(1)

Documents

373753

Sigma-Aldrich

(S)-(−)-Citronellal

96%

Synonym(s):

(−)-Citronellal, (3S)-3,7-Dimethyl-6-octenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CHO
CAS Number:
Molecular Weight:
154.25
Beilstein:
1720790
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

optical activity

[α]20/D −15°, neat

refractive index

n20/D 1.446 (lit.)

density

0.851 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)C[C@@H](C)CC\C=C(\C)C

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,8,10H,4,6-7H2,1-3H3/t10-/m0/s1

InChI key

NEHNMFOYXAPHSD-JTQLQIEISA-N

General description

(S)-(-)-Citronellal is a monoterpenoid compound mainly found in Corymbia citriodora and Cymbopogon nardus essential oils.

Application

(S)-(-)-Citronellal may be used in the synthesis of bioactive compounds like (+)-hexahydrocannabinol, (S)-isopulegol, machaeriols A and B.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient one-pot synthetic approaches for cannabinoid analogues and their application to biologically interesting (-)-hexahydrocannabinol and (+)-hexahydrocannabinol.
Lee YR and Xia L.
Tetrahedron Letters, 49(20), 3283-3287 (2008)
First synthesis of antimalarial Machaeriols A and B.
Chittiboyina AG, et al.
Tetrahedron Letters, 45(8), 1689-1691 (2004)
Asymmetric cyclization of unsaturated aldehydes catalyzed by a chiral Lewis acid.
Sakane S, et al.
Tetrahedron Letters, 26(45), 5535-5538 (1985)
Xiangxian Ying et al.
Molecules (Basel, Switzerland), 24(6) (2019-03-21)
The members of the Old Yellow Enzyme (OYE) family are capable of catalyzing the asymmetric reduction of (E/Z)-citral to (R)-citronellal-a key intermediate in the synthesis of L-menthol. The applications of OYE-mediated biotransformation are usually hampered by its insufficient enantioselectivity and
Alina Mariana Balu et al.
Organic & biomolecular chemistry, 8(12), 2845-2849 (2010-05-01)
The selective conversion of citronellal to menthols, with good diastereoselectivities to (-)-menthol for the case of (+)-citronellal as starting material, can effectively be carried out in a one-step reaction under microwave irradiation catalysed by supported nanoparticles on mesoporous materials. 2%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service