Skip to Content
Merck
All Photos(3)

Key Documents

221198

Sigma-Aldrich

Formamide

ACS reagent, ≥99.5%

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
grade:
ACS reagent
Assay:
≥99.5%
technique(s):
PCR: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

grade

ACS reagent

Quality Level

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

932 °F

technique(s)

PCR: suitable

color

APHA: ≤10

refractive index

n20/D 1.447 (lit.)

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

transition temp

freezing point 2.0-3.0 °C

density

1.134 g/mL at 25 °C (lit.)

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.
The addition of formamide may enhance the specificity and efficiency of amplification of PCR (polymerase chain reaction). It may be used as an ammonia equivalent for the palladium-catalyzed aminocarbonylation of 1-bromo-3-trifluoromethylbenzene to form 3-(trifluoromethyl)benzamide.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formamide can dramatically improve the specificity of PCR.
Sarkar G, et al.
Nucleic Acids Research, 18(24), 7465-7465 (1990)
Weinreb SM.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 21, 126-126 (2014)
Solubilization in formamide protects RNA from degradation.
Chomczynski P.
Nucleic Acids Research, 20(14), 3791-3791 (1992)
J Casey et al.
Nucleic acids research, 4(5), 1539-1552 (1977-01-01)
The thermal stabilities of RNA:DNA hybrids are substantially greater than those of DNA:DNA duplexes in aqueous electrolyte solutions containing high concentrations of formamide. Association rates to form DNA:DNA duplexes and DNA:RNA hybrids have been measured in these solvents. There is
Microwave spectrum and molecular structure of formamide.
Costain CC and Dowling JM.
J. Chem. Phys. , 32(1), 158-165 (1960)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service