Skip to Content
Merck
All Photos(2)

Key Documents

199508

Sigma-Aldrich

Acridine Yellow G

Dye content 90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N3 · HCl
CAS Number:
Molecular Weight:
273.76
Colour Index Number:
46025
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

crystalline powder

Quality Level

composition

Dye content, 90%

solubility

water: 1 mg/mL, clear, yellow to orange

λmax

442 nm

ε (extinction coefficient)

44000 at 262-268 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].Cc1cc2cc3cc(C)c(N)cc3nc2cc1N

InChI

1S/C15H15N3.ClH/c1-8-3-10-5-11-4-9(2)13(17)7-15(11)18-14(10)6-12(8)16;/h3-7H,16-17H2,1-2H3;1H

InChI key

BGLGAKMTYHWWKW-UHFFFAOYSA-N

Biochem/physiol Actions

Acridine Yellow G (AYG) is an organic dye with photophysical properties. It is widely used in optics, medicine and photovoltaic cells. AYG has been used as a solar photocatalyst to increase biodegradability and remove ferulic acid, a target pollutant from wastewater. It retards the growth of glioblastoma via dual inhibition of epidermal growth factor receptor and protein kinase c kinases.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Qi et al.
The Journal of biological chemistry, 287(9), 6113-6127 (2012-01-05)
Amplification of the epidermal growth factor receptor (EGFR), frequently expressed as a constitutively active deletion mutant (EGFRvIII), occurs commonly in glioblastoma multiformes (GBM). However, blockade of EGFR is therapeutically disappointing for gliomas with PTEN deletion. To search for small molecules
T Pérez-Ruíz et al.
Analytical and bioanalytical chemistry, 375(5), 661-665 (2003-03-15)
A flow-injection configuration for the determination of formaldehyde is proposed. The method is based on the enhancing effect of formaldehyde on the oxidation of acridine yellow by bromate in acidic medium. The proposed procedure is simple, inexpensive, sensitive and suitable
M Teuber et al.
Biochimica et biophysica acta, 1506(1), 31-46 (2001-06-22)
Fluorescent DeltapH and DeltaPsi indicators have been screened for the non-invasive monitoring of bioenergetic processes in whole cells of the cyanobacterium Synechocystis sp. PCC 6803. Acridine yellow and Acridine orange proved to be the best DeltapH indicators for the investigation
G J Tangelder et al.
The American journal of physiology, 268(2 Pt 2), H909-H915 (1995-02-01)
A method is presented to assess in vivo in transparent tissues the leukocyte subtypes that roll in microvessels. In nine rabbits anesthetized with ketamine-xylazine, leukocyte nuclei were stained in situ with acridine yellow (3 mg/kg i.v. for 5 min). Intravital
Kęstutis Aidas et al.
The journal of physical chemistry. B, 117(7), 2069-2080 (2013-01-30)
Attempting to unravel mechanisms in optical probing of proteins, we have performed pilot calculations of two cationic chromophores-acridine yellow and proflavin-located at different binding sites within human serum albumin, including the two primary drug binding sites as well as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service