Skip to Content
Merck
All Photos(1)

Key Documents

19780

Sigma-Aldrich

2-Chloro-2-methylpropane

puriss. p.a., ≥99.0% (GC)

Synonym(s):

tert-Butyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCl
CAS Number:
Molecular Weight:
92.57
Beilstein:
1730872
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

3.2 (vs air)

Quality Level

vapor pressure

4.82 psi ( 20 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

evapn. residue

≤0.01%

refractive index

n20/D 1.385 (lit.)
n20/D 1.385

bp

51-52 °C (lit.)

mp

−25 °C (lit.)

density

0.851 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.5 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤2 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

functional group

alkyl halide
chloro

SMILES string

CC(C)(C)Cl

InChI

1S/C4H9Cl/c1-4(2,3)5/h1-3H3

InChI key

NBRKLOOSMBRFMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youngseon Shim et al.
The journal of physical chemistry. B, 112(9), 2637-2643 (2008-02-14)
The S(N)1 ionization reaction RX --> R(+) + X(-) for 2-chloro-2-methylpropane in ionic liquid 1-ethyl-3-methylimidazolium hexafluorophosphate ([emim](+)P(-)(6)) is studied via molecular dynamics computer simulations. By employing a two-state valence-bond description for electronic structure variations of the reaction complex, the free
Maria C Mavroudi et al.
Electrophoresis, 35(18), 2573-2578 (2014-05-24)
The aim of the present study is the CE performance evaluation for the separation of 2-arylpropionic acid nonsteroidal anti-inflammatory drugs. In particular, the separation of indoprofen, carprofen, ketoprofen, ibuprofen, and flurbiprofen was obtained by supporting the BGE either with SDS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service