Skip to Content
Merck
All Photos(1)

Key Documents

17975

Supelco

Ninhydrin reagent according to Stahl

for TLC derivatization

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.22

Quality Level

form

liquid

technique(s)

thin layer chromatography (TLC): suitable

impurities

1-butanol
acetic acid

density

0.823 g/mL at 20 °C

General description

Ninhydrin reagent is mostly used for the detection of amino acids. It basically gives a characteristic purple color on reaction with a-amino acids and other amino compounds, provided the pH of the reaction is 5.0 or higher. At an acid pH of 0.3, the reaction of ninhydrin with ornithinine and proline results in a red color product. Ninhydrin reagent can stay stable at room temperature in the absence of nitrogen atmosphere for upto 1 month in the ethylene glycol-sodium acetate solvent system.

Application

1 g ninhydrin dissolved in 475 mL 1-butanol and 25 mL acetic acid
Ninhydrin reagent may be used for the quantitative determination of total protein content in tissue hydrolysates.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Starcher
Analytical biochemistry, 292(1), 125-129 (2001-04-26)
Quantitation of small tissue samples for total protein content is essential for many biochemical analyses. In this study a ninhydrin method for measuring the total protein content of tissue hydrolysates is presented. The ninhydrin reagent is stable at room temperature
M K Gaitonde
The Biochemical journal, 104(2), 627-633 (1967-08-01)
1. An acid ninhydrin reagent was found to react specifically in forming a pink product (E(max.) 560mmu) with cysteine. 2. The method was highly sensitive for the determination of cysteine (in 28.0x10(3)). Homocysteine, glutathione, proline, ornithine and other naturally occurring
Thin Layer Chromatography in Phytochemistry.
Hajnos WM
Science, 312-312 (2008)
Sylvia Kristyanto et al.
Journal of microbiology (Seoul, Korea), 56(5), 317-323 (2018-05-04)
A novel, aerobic, Gram-stain-negative, non-motile, non-spore forming, rod-shaped bacterium, designated strain Dol 15-39T, was isolated from a seawater sample near Geoje Island in the South Sea, Republic of Korea. The strain was found to be oxidase-negative and catalase-positive. The isolate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service