Skip to Content
Merck
All Photos(1)

Key Documents

15716

Supelco

Boron trifluoride-methanol solution

~10% (~1.3 M), for GC derivatization, LiChropur

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · MeOH
CAS Number:
Molecular Weight:
99.85
Beilstein:
3611499
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.05

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

concentration

~10% (~1.3 M)

technique(s)

gas chromatography (GC): suitable

density

0.86 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CO.FB(F)F

InChI

1S/CH4O.BF3/c1-2;2-1(3)4/h2H,1H3;

InChI key

JBXYCUKPDAAYAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boron trifluoride (BF3) is a Lewis acid; it is a very useful reagent for forming esters from acids. Boron trifluoride is commercially available as a 14% solution in methanol for forming esters derivatives. It catalysed reactions carried out in a water bath are very rapid and can be completed in a few minutes. Boron-trifluoride-catalysed transesterification of vegetable oils is one of the most popular methods. For transesterification of vegetable oils, BF3 is used in methanol (15 to 20% w/v).

Application

Learn more in the Product Information
Boron trifluoride-methanol solution may be used for catalysing a method, used for the synthesis of bishydroperoxides and their derivatives, based on a reaction of ketals with a solution of hydrogen peroxide in anhydrous ether. It may also be used for the rapid preparation of fatty acid esters for gas chromatographic analysis, as it converts fatty acids to their respective methyl esters in about 2 minutes.
Suitable for derivatization of hydroxy fatty acids.

Other Notes

Reagent for esterifying fatty acid for gas chromatographic analysiscitation
Reagent for methyl ester, O-acetyl.
Reagent for esterifying fatty acid for gas chromatographic analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 1

Target Organs

Eyes,Central nervous system, Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodiesel: A Realistic Fuel Alternative for Diesel Engines.
Demirbas A.
Technology & Engineering, 123-124 (2007)
The rapid preparation of fatty acid esters for gas chromatographic analysis.
Metcalfe LD and Schmitz AA.
Analytical Chemistry, 33 (3), 363-364 (1961)
A new method for the synthesis of bishydroperoxides based on a reaction of ketals with hydrogen peroxide catalyzed by boron trifluoride complexes.
Alexander TO
Tetrahedron Letters, 44 (39), 7359-7363 (2003)
Christian F Ruiz et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(8), 10574-10589 (2020-06-23)
Cancer cells require extensive metabolic reprograming in order to provide the bioenergetics and macromolecular precursors needed to sustain a malignant phenotype. Mutant KRAS is a driver oncogene that is well-known for its ability to regulate the ERK and PI3K signaling
H. Rotzsche
Journal of Chromatography A, 552, 281-281 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service