Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boron trifluoride-methanol solution
Linear Formula:
BF
3
· MeOH
CAS Number:
373-57-9
Molecular Weight:
99.85
EC Number:
206-766-4
Products
Product Category
acids (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Functional Group
Reaction Type
Reagent Type
Core
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
B1252
Boron trifluoride-methanol solution,
14% in methanol
Expand
View Pricing
134821
Boron trifluoride-methanol solution,
50% w/w in methanol
Expand
View Pricing
15716
Boron trifluoride-methanol solution,
~10% (~1.3 M), for GC derivatization, LiChropur
™
Expand
View Pricing