Skip to Content
Merck
All Photos(1)

Key Documents

14795

Sigma-Aldrich

N,N,N′,N′-Tetramethyl-1,8-naphthalenediamine

purum, ≥99.0% (NT)

Synonym(s):

1,8-Bis(dimethylamino)naphthalene, DMAN, Proton-sponge®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2
CAS Number:
Molecular Weight:
214.31
Beilstein:
396782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥99.0% (NT)

form

solid

mp

45-49 °C

functional group

amine

SMILES string

CN(C)c1cccc2cccc(N(C)C)c12

InChI

1S/C14H18N2/c1-15(2)12-9-5-7-11-8-6-10-13(14(11)12)16(3)4/h5-10H,1-4H3

InChI key

GJFNRSDCSTVPCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N,N′,N′-Tetramethyl-1,8-naphthalenediamine was used in protonation of multiple-charged oligonucleotide anions. It was used as reagent during reaction of benzaldehyde with acetic anhydride catalyzed by bismuth nitrate. It was used as extractant during separation of organic acids from fermentation broths by liquid-liquid extraction.

Other Notes

Strong amine base ("proton sponge") with unusual properties

Legal Information

Proton-sponge is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bismuth compounds in organic synthesis. Bismuth nitrate catalyzed chemoselective synthesis of acylals from aromatic aldehydes.
Aggen DH, et al.
Tetrahedron, 60(16), 3675-3679 (2004)
Extractant screening for liquid-liquid extraction in environmentally benign production routes.
Krzyzaniak A, et al.
Chemical Engineering Transactions, 24, 709-714 (2011)
R.L. Benoit et al.
Canadian Journal of Chemistry, 65, 996-996 (1987)
Yu Xia et al.
Analytical chemistry, 77(11), 3683-3689 (2005-06-01)
A single sonic spray source has been used to generate both positive and negative ions for subsequent ion/ion reaction experiments. Ion/ion reactions took place after ions of each polarity were sequentially injected into a linear ion trap, where axial trapping
R.W. Alder et al.
Journal of the Chemical Society. Chemical Communications, 723-723 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service