Skip to Content
Merck
All Photos(3)

Key Documents

125660

Sigma-Aldrich

Leucomalachite Green

powder

Synonym(s):

4,4′-Benzylidenebis(N,N-dimethylaniline)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[C6H4N(CH3)2]2
CAS Number:
Molecular Weight:
330.47
Beilstein:
2140506
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

technique(s)

microbe id | staining: suitable

mp

100-102 °C (lit.)

λmax

623 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)C(c2ccccc2)c3ccc(cc3)N(C)C

InChI

1S/C23H26N2/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,23H,1-4H3

InChI key

WZKXBGJNNCGHIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Leucomalachite green has been used as a reagent in presumptive tests which are used to detect invisible blood stains.

Biochem/physiol Actions

Leucomalachite green (LMG) is a chief metabolite of malachite green, a triphenylmethane dye that has been used generally as an antifungal drug in the fish industry. DNA adducts formed in livers of rats fed with leucomalachite green have been observed with little mutagenic or carcinogenic effect. LMG is one of the most commonly used reagents for presumptive blood test. It gives a characteristic green color in the presence of blood. LMG destroys DNA in the blood, therefore, the chances of DNA recovery and amplification is reduced or eliminated.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High incidence of blood exposure due to imperceptible contaminated splatters during oral surgery.
Ishihama K
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 66(4), 704-710 (2008)
Mutagenicity and carcinogenicity in relation to DNA adduct formation in rats fed leucomalachite green.
Culp SJ
Mutation Research, 506-507, 55-63 (2002)
Evidence of aerosolised floating blood mist during oral surgery.
Ishihama K
Journal of Hospital Infection, 71(4), 359-364 (2009)
Zoe Hall et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 874(1-2), 95-100 (2008-09-27)
A high accuracy method for the quantification of malachite green (MG) and leucomalachite green (LMG) in salmon is described. Analytical challenges including the effects of analyte instability and matrix suppression were minimised by the use of exact matching isotope dilution
Chaoyang Long et al.
Journal of chromatography. A, 1203(1), 21-26 (2008-07-29)
With a new oxidant for post-column chemical derivation, a novel approach was developed for the determination of Malachite Green (MG) and Leucomalachite Green (LMG) in fish by high-performance liquid chromatography (HPLC). A C(8) column was used for separation, and elution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service