Skip to Content
Merck
All Photos(1)

Documents

11536

Sigma-Aldrich

Lead(II,IV) oxide

≥96%

Synonym(s):

Minium, Red lead oxide, Trilead tetraoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb3O4
CAS Number:
Molecular Weight:
685.60
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

10 mmHg ( 0 °C)

Quality Level

Assay

≥96%

form

solid

reaction suitability

core: lead
reagent type: catalyst

impurities

≤0.1% in HNO3/H2O2 insoluble matter

anion traces

chloride (Cl-): ≤100 mg/kg

SMILES string

O1[PbH2]O[Pb]12O[PbH2]O2

InChI

1S/4O.3Pb

InChI key

XMFOQHDPRMAJNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lead(II,IV) oxide is a red pigment commonly used as an anti-corrosive pigment in protective coatings and as electrodes in batteries. In addition, it is also used as a catalyst for the Diels-Alder cycloaddition and as an oxidant for the generation of quinones from hydroquinones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Ox. Sol. 2 - Repr. 1A - STOT RE 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical Perforance and Synthesis of Nanostructured Lead Oxide
Wang H
Modern applied science, 4(4), 116-116 (2010)
Synthesis of Six-Membered Functionalized Carbocyclic Compounds by One-Pot Reaction of Hydroquinone Derivatives and Dienes
Cai GR, et al.
Synthetic Communications, 41(20), 3016-3025 (2011)
Guo-zhen Lin et al.
Clinica chimica acta; international journal of clinical chemistry, 413(13-14), 1156-1159 (2012-05-09)
Traditional medicines or ethnic remedies, such as Ayurvedic drug of India, have been reported in numerous cases to be one of the main exposure risks for severe lead poisoning. We describe the poisoning of 2 young children from a Chinese
Yangen Zhou et al.
Inorganic chemistry, 51(23), 12594-12596 (2012-11-13)
Pb(3)O(4) could react with organic dyes in aqueous solution under visible light irradiation, in which Pb(3)O(4) was transformed into Pb(3)(CO(3))(2)(OH)(2) along with oxidation of the organic dyes. Cu(2+) has considerable effect on the reaction. In the presence of Cu(2+), MO
A Fischbein et al.
JAMA, 247(14), 2007-2009 (1982-04-09)
A case of lead poisoning in a female art conservator is reported. The patient had experienced excessive lead exposure while restoring an antique Peruvian tapestry from the Chancay period (1000 to 1500 AD) using a powdered pigment (cinnabar), which had

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service