Skip to Content
Merck
All Photos(3)

Key Documents

307734

Sigma-Aldrich

Lead(II) sulfate

98%

Synonym(s):

Anglesite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbSO4
CAS Number:
Molecular Weight:
303.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
powder and chunks
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

powder and chunks

reaction suitability

reagent type: catalyst
core: lead

density

6.2 g/mL at 25 °C (lit.)

SMILES string

[PbH2++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.Pb/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2

InChI key

KEQXNNJHMWSZHK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Falgayrac et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(5), 1095-1101 (2006-03-10)
Currently, the smelting activities of lead and zinc are the loudest sources of local pollution by emission in the troposphere of dust of micrometer size containing PbSO(4). As the particles evolve in the troposphere, their chemical and physical properties -
K R Logner et al.
Journal of dairy science, 67(5), 1007-1013 (1984-05-01)
Sixteen Holstein intact male calves averaging 85 kg and 74 days of age were assigned randomly to four dietary lead treatments according to body weight. They were fed for ad libitum consumption a control diet containing no added lead or
Martina Vítková et al.
Journal of hazardous materials, 167(1-3), 427-433 (2009-02-07)
The leaching behaviour of fly ash (FA) from a secondary Pb smelter was assessed using the pH-static leaching experiment according to prEN 14997 (pH range 3-11) coupled with mineralogical investigation of the leached FA by XRD and Rietveld analyses and
Byung-Ryool Hyun et al.
Nano letters, 11(5), 2126-2132 (2011-04-22)
The roles of solvent reorganization energy and electronic coupling strength on the transfer of photoexcited electrons from PbS nanocrystals to TiO(2) nanoparticles are investigated. We find that the electron transfer depends only weakly on the solvent, in contrast to the
J J Kazama et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 69(6), 324-328 (1994-11-01)
We describe a new technique for immunohistochemical and enzyme-histochemical double staining using confocal laser scanning microscopy in the reflection mode. As an example, we investigated the immunoreactivity for Spot 35-calbindin-D28K, a vitamin D-dependent calcium binding protein, and the enzyme activity

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service