Skip to Content
Merck
All Photos(1)

Documents

30-1220

Sigma-Aldrich

N,N,N′,N′-Tetramethylethylenediamine

≥99.0%

Synonym(s):

1,2-Bis(dimethylamino)ethane, TEMED, TMEDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1732991
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥99.0%

form

liquid

expl. lim.

9.08 %

availability

available only in Japan

refractive index

n20/D 1.4179 (lit.)

bp

120-122 °C (lit.)

mp

−55 °C (lit.)

density

0.775 g/mL at 20 °C (lit.)

SMILES string

CN(C)CCN(C)C

InChI

1S/C6H16N2/c1-7(2)5-6-8(3)4/h5-6H2,1-4H3

InChI key

KWYHDKDOAIKMQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

for electrophoresis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

61.7 °F - closed cup

Flash Point(C)

16.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jack K Clegg et al.
Dalton transactions (Cambridge, England : 2003), 39(11), 2804-2815 (2010-03-05)
New examples of nitrogen base adducts of dinuclear Co(II), Ni(II) and Cu(II) complexes of the doubly deprotonated forms of 1,3-aryl linked bis-beta-diketones of type [RC(=O)CH(2)C(=O)C(6)H(4)C(=O)CH(2)C(=O)R] (L(1)H(2)) incorporating the mono- and difunctional amine bases pyridine (Py), 4-ethylpyridine (EtPy), piperidine (pipi), 1,4-piperazine
Kazuhiro Uehara et al.
Inorganic chemistry, 49(4), 2008-2015 (2010-01-26)
Reactions of (en*)Pd(NO(3))(2) (en* = N,N,N',N'-tetramethylethylenediamine)with a series of organic bridging ligands of pyrazine, 1,2-bis(4-pyridyl)ethylene, 1,2-bis(4-pyridyl)acetylene, and 1,4-bis(4-pyridyl)benzene are carried out to investigate factors controlling the supramolecular structures and the equilibrium between the molecular triangles and the squares in solutions.
Kiyohito Shimura
Electrophoresis, 30(1), 11-28 (2008-12-25)
The methodological advancements in CIEF in tubes and microchips during the period between 2002 and early 2008 are reviewed as a continuation of previous review by the same author (Electrophoresis 2002, 23, 3847-3857). After a brief introduction, the following topics
Abu Bin Imran et al.
Nature communications, 5, 5124-5124 (2014-10-09)
Stimuli-sensitive hydrogels changing their volumes and shapes in response to various stimulations have potential applications in multiple fields. However, these hydrogels have not yet been commercialized due to some problems that need to be overcome. One of the most significant
Andrew J Ross et al.
The Journal of organic chemistry, 76(6), 1727-1734 (2011-02-16)
The structure of the alkylzinc-tetramethylethyl-enediamine (TMEDA) cluster cation 3 has been determined in the gas phase by a combination of tandem mass spectrometry, infrared multiphoton dissociation (IRMPD) spectroscopy, and DFT calculations. Both sets of experimental results establish the existence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service