Skip to Content
Merck
All Photos(1)

Key Documents

30-0610

Sigma-Aldrich

1,1,2,2-Tetrachloroethane

JIS special grade, ≥97.0%

Synonym(s):

Acetylene tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHCl2CHCl2
CAS Number:
Molecular Weight:
167.85
Beilstein:
969206
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
JIS special grade
bp:
147 °C (lit.)
vapor pressure:
8 mmHg ( 20 °C)

grade

JIS special grade

vapor density

5.8 (vs air)

vapor pressure

8 mmHg ( 20 °C)

Assay

≥97.0%

form

liquid

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.494 (lit.)

bp

147 °C (lit.)

mp

−43 °C (lit.)

density

1.586 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)C(Cl)Cl

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H

InChI key

QPFMBZIOSGYJDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Srigokul Upadhyayula et al.
The journal of physical chemistry. B, 115(30), 9473-9490 (2011-06-21)
Electrostatic properties of proteins are crucial for their functionality. Carboxyamides are small polar groups that, as peptide bonds, are principal structural components of proteins that govern their electrostatic properties. We investigated the medium dependence of the molar polarization and of
Debolina Basu et al.
Bioresource technology, 101(1), 21-25 (2009-08-25)
This study examines the performance of bench-scale Upflow Anaerobic Sludge Blanket (UASB) reactor in treating simulated wastewaters containing 1,1,2,2-tetrachloroethane (TeCA). Reactors R1 (control) and R2 (containing TeCA) were operated at Hydraulic Retention Times (HRTs) of 36, 30, 24, 18 and
Marcos Caroli Rezende et al.
Magnetic resonance in chemistry : MRC, 47(6), 505-510 (2009-04-01)
Spectra of the N-phenyl-5,6-dihydro-2,4-diphenylbenzo[h]quinolinium tetrafluoroborate (1) and of the N-phenyl-5,6,8,9-tetrahydro-7-phenyldibenzo[c,h]acridinium tetrafluoroborate (2) were recorded in various solvents and temperatures. The analysis of the (1)H-NMR spectra of the tetrafluoroborate salt 1, recorded in acetone, acetonitrile, 1,1,2,2-tetrachloroethane and chloroform, revealed the existence
Wan-Chun Tsai et al.
Journal of chromatography. A, 1216(45), 7846-7850 (2009-10-07)
A novel sample preparation method "Dispersive liquid-liquid-liquid microextraction" (DLLLME) was developed in this study. DLLLME was combined with liquid chromatography system to determine chlorophenoxy acid herbicide in aqueous samples. DLLLME is a rapid and environmentally friendly sample pretreatment method. In
Simona Rossetti et al.
Biotechnology and bioengineering, 100(2), 240-249 (2008-04-24)
An anaerobic microcosm set up with aquifer material from a 1,1,2,2-tetrachloroethane (TeCA) contaminated site and amended with butyrate showed a complete TeCA dechlorination to ethene. A structure analysis of the microbial community was performed by fluorescence in situ hybridization (FISH)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service