Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,1,2,2-Tetrachloroethane
Synonym(s):
Acetylene tetrachloride, 1,1,2,2-Tetrachloroethane
Linear Formula:
CHCl
2
CHCl
2
CAS Number:
79-34-5
Molecular Weight:
167.85
EC Number:
201-197-8
Products
Product Category
solvents (2)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
185434
1,1,2,2-Tetrachloroethane,
reagent grade, ≥98%
Expand
View Pricing
8.43948
1,1,2,2-Tetrachloroethane,
for synthesis
Expand
View Pricing
30-0610
1,1,2,2-Tetrachloroethane,
JIS special grade, ≥97.0%
Expand
View Pricing
46259
1,1,2,2-Tetrachloroethane,
analytical standard
Expand
View Pricing