Skip to Content
Merck
All Photos(1)

Key Documents

30-0460

Sigma-Aldrich

1,1,2,2-Tetrabromoethane

SAJ first grade, ≥98.0%

Synonym(s):

Acetylene tetrabromide, Muthmanns liquid, TBE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2CHCHBr2
CAS Number:
Molecular Weight:
345.65
Beilstein:
1098321
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
SAJ first grade
Assay:
≥98.0%
bp:
119 °C/15 mmHg (lit.)
vapor pressure:
0.1 mmHg ( 20 °C)

grade

SAJ first grade

vapor density

11.9 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

635 °F

availability

available only in Japan

refractive index

n20/D 1.637 (lit.)

bp

119 °C/15 mmHg (lit.)

mp

−1-1 °C (lit.)

density

2.967 g/mL at 25 °C (lit.)

SMILES string

BrC(Br)C(Br)Br

InChI

1S/C2H2Br4/c3-1(4)2(5)6/h1-2H

InChI key

QXSZNDIIPUOQMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Galuppi et al.
Veterinary parasitology, 210(1-2), 10-18 (2015-04-15)
The present study aims to evaluate the prevalence, pattern of spread and risk factors for the transmission of cryptosporidiosis in foals and mares hospitalized in a University Equine Perinatology Unit, where a new subtype family of Cryptosporidium horse genotype was
Zhaojiang Guo et al.
Journal of invertebrate pathology, 126, 21-30 (2015-01-18)
The Gram-positive bacterium Bacillus thuringiensis (Bt) produces Cry toxins that have been used to control important agricultural pests. Evolution of resistance in target pests threatens the effectiveness of these toxins when used either in sprayed biopesticides or in Bt transgenic
Anusha M Gopalakrishnan et al.
Antimicrobial agents and chemotherapy, 59(1), 317-325 (2014-10-29)
Artemisinin-based combination therapy (ACT) is the recommended first-line treatment for Plasmodium falciparum malaria. It has been suggested that the cytotoxic effect of artemisinin is mediated by free radicals followed by the alkylation of P. falciparum proteins. The endoperoxide bridge, the
Stefania Napolitano et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(13), 2975-2983 (2015-04-04)
In colorectal cancer, the activation of the intracellular RAS-RAF and PIK3CA-AKT pathways has been implicated in the resistance to anti-EGFR mAbs. We have investigated the role of regorafenib, an oral multikinase inhibitor, in combination with cetuximab, an anti-EGFR mAb, to
Somin Eunice Lee et al.
Nano letters, 15(7), 4564-4570 (2015-06-04)
Plasmon rulers, consisting of pairs of gold nanoparticles, allow single-molecule analysis without photobleaching or blinking; however, current plasmon rulers are irreversible, restricting detection to only single events. Here, we present a reversible plasmon ruler, comprised of coupled gold nanoparticles linked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service