Skip to Content
Merck
1,1,2,2-Tetrabromoethane SAJ first grade, ≥98.0%

1,1,2,2-Tetrabromoethane

1,1,2,2-Tetrabromoethane SAJ first grade, ≥98.0%
Synonym(s):
Acetylene tetrabromide, Muthmanns liquid, TBE, 1,1,2,2-Tetrabromoethane
Linear Formula:
Br2CHCHBr2
CAS Number:
Molecular Weight:
345.65

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Special Grade