Skip to Content
Merck
All Photos(1)

Documents

28-3730

Sigma-Aldrich

Sodium phosphate dibasic dodecahydrate

SAJ first grade, ≥98.0

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 12H2O
CAS Number:
Molecular Weight:
358.14
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.0

form

crystalline

availability

available only in Japan

pH

8.4-9.6 (25 °C, 35.8 g/L)

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.12H2O/c;;1-5(2,3)4;;;;;;;;;;;;/h;;(H3,1,2,3,4);12*1H2/q2*+1;;;;;;;;;;;;;/p-2

InChI key

DGLRDKLJZLEJCY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew H Crouthamel et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(11), 2625-2632 (2013-08-24)
Elevated serum phosphate has emerged as a major risk factor for vascular calcification. The sodium-dependent phosphate cotransporter, PiT-1, was previously shown to be required for phosphate-induced osteogenic differentiation and calcification of cultured human vascular smooth muscle cells (VSMCs), but its
Matthew R Weir et al.
Journal of clinical hypertension (Greenwich, Conn.), 16(12), 875-882 (2014-10-21)
The effects of canagliflozin, a sodium glucose co-transporter 2 inhibitor, on blood pressure (BP) and osmotic diuresis- and intravascular volume reduction-related adverse events (AEs) were evaluated using pooled data from four placebo-controlled, phase 3 studies in patients with type 2
George G S Sandor et al.
International journal of cardiology, 179, 470-475 (2014-12-04)
Patients with Marfan (MFS) and Loeys-Dietz (LDS) syndromes have been shown to have abnormal aortic biophysical properties. The purpose of this study was to compare the effects of 12-months of therapy with atenolol or losartan on vascular function in young
Luciano Candilio et al.
Heart (British Cardiac Society), 101(3), 185-192 (2014-09-26)
Remote ischaemic preconditioning (RIPC), using brief cycles of limb ischaemia/reperfusion, is a non-invasive, low-cost intervention that may reduce perioperative myocardial injury (PMI) in patients undergoing cardiac surgery. We investigated whether RIPC can also improve short-term clinical outcomes. One hundred and
Karen Sliwa et al.
Heart (British Cardiac Society), 100(24), 1967-1974 (2014-09-18)
Lack of evidence-based data on the spectrum of cardiovascular disease (CVD) in pregnancy or in the postpartum period, as well as on maternal and fetal outcome, provides challenges for treating physicians, particularly in areas of low resources. The objectives of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service