Skip to Content
Merck
All Photos(1)

Key Documents

28-2940

Sigma-Aldrich

Sodium hydroxide

JIS special grade, ≥96.0%

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

JIS special grade

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

Assay

≥96.0%

form

pellets

availability

available only in Japan

mp

318 °C (lit.)

solubility

water: ~1260 g/L at 20 °C

density

2.13 g/cm3

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Facile Synthesis of Carbon Dots from Amido Black 10b for Sensing in Real Samples.: This study presents a simple synthesis method for carbon dots using Amido Black 10b, with applications in sensing. The researchers demonstrated the utility of these carbon dots in real sample analysis, highlighting their potential in environmental monitoring and biomedical applications. (Li et al., 2022).

  • Multiple chemical modifications and Cd(2+) adsorption characteristics of sludge-based activated carbon.: The research explores various chemical modifications of sludge-based activated carbon and its effectiveness in adsorbing cadmium ions (Cd(2+)). Sodium hydroxide was used in the modification process, enhancing the material′s adsorption capabilities. This study is relevant for environmental cleanup and water treatment applications. (Chen et al., 2022).

  • Characterization of ligno-cellulosic fiber extracted from Atriplex halimus L. plant.: This study characterizes ligno-cellulosic fibers extracted from Atriplex halimus using sodium hydroxide. The results show the potential of these fibers in biocomposites, contributing to sustainable material development. (Belouadah et al., 2021).

  • Monitoring the extraction of copper from chicken dung leachate using an aluminium electrode as an indicator.: The research focuses on the extraction of copper from chicken dung leachate, using an aluminium electrode for monitoring. Sodium hydroxide is employed in the leachate treatment process, showcasing its role in resource recovery from agricultural waste. (Kugeria et al., 2019).

  • Coconut coir pith lignin: A physicochemical and thermal characterization.: This paper discusses the characterization of lignin from coconut coir pith, using sodium hydroxide in the extraction process. The findings are significant for the development of bio-based materials and the utilization of agricultural residues. (Panamgama et al., 2018).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Virginie Mariot et al.
Annals of neurology, 78(3), 387-400 (2015-05-29)
Facioscapulohumeral muscular dystrophy (FSHD) is linked to either contraction of D4Z4 repeats on chromosome 4 or to mutations in the SMCHD1 gene, both of which result in the aberrant expression of the transcription factor DUX4. However, it is still difficult
Konstantinos A Polyzos et al.
Cardiovascular research, 106(2), 295-302 (2015-03-10)
Atherosclerosis is a chronic inflammatory disease that is initiated by the retention and accumulation of low-density lipoprotein in the artery, leading to maladaptive response of cells from the immune system and vessel wall. Strong evidence implicates indoleamine 2,3-dioxygenase (IDO), the
C Berger et al.
Human reproduction (Oxford, England), 30(4), 800-811 (2015-03-06)
Does ulipristal acetate (UPA) used for emergency contraception (EC) interfere with the human embryo implantation process? UPA, at the dosage used for EC, does not affect human embryo implantation process, in vitro. A single pre-ovulatory dose of UPA (30 mg)
Valdeir Arantes et al.
Biotechnology for biofuels, 7, 87-87 (2014-07-01)
There is still considerable debate regarding the actual mechanism by which a "cellulase mixture" deconstructs cellulosic materials, with accessibility to the substrate at the microscopic level being one of the major restrictions that limits fast, complete cellulose hydrolysis. In the
Nicole von Burg et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(35), 12835-12840 (2014-08-20)
Group 3 innate lymphoid cells (ILC3s) have emerged as important cellular players in tissue repair and innate immunity. Whether these cells meaningfully regulate adaptive immune responses upon activation has yet to be explored. Here we show that upon IL-1β stimulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service