Skip to Content
Merck
All Photos(1)

Key Documents

24-6470

Sigma-Aldrich

Pyoktanin blue solution

1 % (w/v)

Synonym(s):

Basic violet 1 solution, Methyl violet 2B solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

form

liquid

availability

available only in Japan

concentration

1 % (w/v)

dilution

(for analytical testing)

Suitability

bacterial staining

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service