Skip to Content
Merck
All Photos(1)

Key Documents

24-0990

Sigma-Aldrich

Peracetic acid solution

8.7% in acetic acid

Synonym(s):

Peroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO3H
CAS Number:
Molecular Weight:
76.05
Beilstein:
1098464
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

form

liquid

reaction suitability

reagent type: oxidant

availability

available only in Japan

concentration

8.7% in acetic acid

storage temp.

−20°C

SMILES string

CC(=O)OO

InChI

1S/C2H4O3/c1-2(3)5-4/h4H,1H3

InChI key

KFSLWBXXFJQRDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

102.2 °F - acetic acid

Flash Point(C)

39 °C - acetic acid


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinqiang Xie et al.
Nucleic acids research, 40(18), 9115-9124 (2012-07-10)
Diverse bacteria contain DNA with sulfur incorporated stereo-specifically into their DNA backbone at specific sequences (phosphorothioation). We found that in vitro oxidation of phosphorothioate (PT) DNA by hydrogen peroxide (H(2)O(2)) or peracetic acid has two possible outcomes: DNA backbone cleavage
G Hey et al.
Environmental technology, 33(7-9), 1041-1047 (2012-06-23)
Removal of six active pharmaceutical ingredients in wastewater was investigated using chlorine dioxide (ClO2) or peracetic acid (PAA) as chemical oxidants. Four non-steroidal anti-inflammatory drugs (ibuprofen, naproxen, diclofenac and mefenamic acid) and two lipid-regulating agents (gemfibrozil and clofibric acid, a
Hanem F Khater et al.
Parasitology research, 112(1), 259-269 (2012-10-24)
The fowl tick, Argas persicus (Oken), is of veterinary importance as a parasite of poultry and wild birds. The antitick efficacy, in vitro and in vivo, of peracetic acid (PAA) and deltamethrin (DMT) was tested separately against A. persicus through
Abelardo González et al.
Water environment research : a research publication of the Water Environment Federation, 84(3), 247-253 (2012-07-05)
Disinfection of an advanced primary effluent using a continuous-flow combined peracetic acid/ultraviolet (PAA/UV) radiation system was evaluated. The purpose was to determine whether the maximum microbial content, established under Mexican standards for treated wastewaters meant for reuse--less than 240 most
Beatrice Blanc et al.
Dalton transactions (Cambridge, England : 2003), 42(9), 3156-3169 (2012-12-18)
The chlorite dismutases (Clds) degrade ClO(2)(-) to O(2) and Cl(-) in perchlorate respiring bacteria, and they serve still poorly defined cellular roles in other diverse microbes. These proteins share 3 highly conserved Trp residues, W155, W156, and W227, on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service