Skip to Content
Merck
All Photos(1)

Key Documents

1.07222

Supelco

Peracetic acid

about 38-40%

Synonym(s):

Ethaneperoxoic acid, Peroxyacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.21

Product Name

Peracetic acid about 38-40%,

vapor pressure

14 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

225 °C

potency

263 mg/kg LD50, oral (Rat)

concentration

37.5-40.5% (peracetic acid)

pH

1 (20 °C in H2O)

mp

-37 °C

transition temp

flash point 62 °C

density

1.14 g/cm3 at 20 °C

storage temp.

15-25°C

Related Categories

Application


  • Development of the radiosynthesis of high-specific-activity 123I-NKJ64.: This study by Tavares et al. (2011) presents the development and optimization of the radiosynthesis of high-specific-activity 123I-NKJ64, a compound used in nuclear medicine. The synthesis process involved the use of 38% peracetic acid as a key reagent to achieve the desired radioisotope incorporation and purity. This research highlights the importance of peracetic acid in the production of radiopharmaceuticals, demonstrating its role in enhancing the efficiency and effectiveness of radiosynthetic procedures (Tavares et al., 2011).

Analysis Note

Assay (peracetic acid)37.5 - 40.5 %Identity (IR-spectrum)passes test

Date of expiry: see product label

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Org. Perox. D - Self-react. D - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 2

Flash Point(F)

143.6 °F

Flash Point(C)

62 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service