Skip to Content
Merck
All Photos(1)

Key Documents

15-1490

Sigma-Aldrich

Iron(II) sulfate heptahydrate

JIS special grade, 99.0-102.0%

Synonym(s):

Ferrous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
JIS special grade
form:
solid

grade

JIS special grade

vapor pressure

14.6 mmHg ( 25 °C)

Assay

99.0-102.0%

form

solid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

3.0-4.0 (25 °C, 50 g/L)

density

1.898 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanyan Wei et al.
Food chemistry, 138(2-3), 1952-1958 (2013-02-16)
The present study evaluated the effectiveness of germination and iron fortification on iron concentration and bioavailability of brown rice. Iron fortification during germination process with 0.05-2 g/L ferrous sulfate increased the iron concentration in germinated brown rice from 1.1 to
Huifang Wu et al.
Journal of hazardous materials, 243, 86-94 (2012-11-13)
An experiment was conducted in a batch reactor for a real printing and dyeing wastewater pretreatment using Fenton process in this study. The results showed that original pH, hydrogen peroxide concentration and ferrous sulfate concentration affected ORP value and pretreatment
Roshini Ebenezer et al.
Tropical medicine & international health : TM & IH, 18(8), 942-951 (2013-07-11)
To assess the impact of deworming and iron supplementation on the cognitive abilities and educational achievement of school-age children in Sri Lanka. Prospective, placebo-controlled randomised study. The treatment group received deworming and weekly iron supplementation for 6 months; the control group
Hsiao-Chi Chuang et al.
Toxicology and applied pharmacology, 266(3), 329-334 (2012-12-25)
Pulmonary epithelial lining fluid (ELF) is the first substance to make contact with inhaled particulate matter (PM) and interacts chemically with PM components. The objective of this study was to determine the role of ELF in oxidative stress, DNA damage
Xiao-Kui Ma et al.
Carbohydrate polymers, 92(2), 1188-1196 (2013-02-13)
The influence of several components on exopolysaccharide (EPS) production and antioxidative activity (TEAC, Trolox-Equivalent Antioxidant Capacity) as well as their effects on the morphological development and cell viability of Phellinus sp. P0988 was determined. Rutin, FeSO(4), Tween 80 and complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service