Skip to Content
Merck
All Photos(1)

Key Documents

03-4881

Sigma-Aldrich

4-tert-Butylcatechol

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3-1,2-(OH)2
CAS Number:
Molecular Weight:
166.22
Beilstein:
2043335
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.0%

availability

available only in Japan

bp

285 °C (lit.)

mp

52-55 °C (lit.)

SMILES string

CC(C)(C)c1ccc(O)c(O)c1

InChI

1S/C10H14O2/c1-10(2,3)7-4-5-8(11)9(12)6-7/h4-6,11-12H,1-3H3

InChI key

XESZUVZBAMCAEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martín A Fernández-Baldo et al.
Talanta, 79(3), 681-686 (2009-07-07)
Botrytis cinerea is a plant-pathogenic fungus that produces the disease known as grey mould in a wide variety of agriculturally important hosts in many countries. This paper describes the development of an immunosensor coupled to carbon-based screen-printed electrodes (SPCE) modified
Contact sensitivity to para-tertiary-butylcatechol in an artificial limb.
A W Macfarlane et al.
Contact dermatitis, 22(1), 56-57 (1990-01-01)
T Kawashima et al.
The Journal of investigative dermatology, 82(1), 53-56 (1984-01-01)
4-Tertiary butyl catechol (TBC) causes depigmentation in humans and animals and stimulates formation of pheomelanosomes. In this study, we investigated the effects of noncytotoxic doses of TBC on glutathione S-transferase (GST) activity in the skin of Uscd strain mice and
An unusual delayed patch test reaction to para-tertiary-butylcatechol.
R C Yu et al.
Contact dermatitis, 22(2), 110-111 (1990-02-01)
J N Rodríguez-López et al.
Biochimica et biophysica acta, 1548(2), 238-256 (2001-08-22)
Using gas chromatography-mass spectrometry, the direct enzymatic release of o-diphenol (4-tert-butylcatechol) during the action of tyrosinase on a monophenol (4-tert-butylphenol) has been demonstrated for the first time in the literature. The findings confirm the previously proposed mechanism to explain the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service