Skip to Content
Merck
All Photos(1)

Documents

03-2710

Sigma-Aldrich

Bispyrazolone

JIS special grade, ≥97.0%

Synonym(s):

3,3′-Dimethyl-1,1′-diphenyl-(4,4′-bi-2-pyrazoline)-5,5′-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18N4O2
CAS Number:
Molecular Weight:
346.38
Beilstein:
572787
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

JIS special grade

Assay

≥97.0%

form

powder

availability

available only in Japan

SMILES string

CC1=NN(c2ccccc2)C(=O)C1C3C(C)=NN(c4ccccc4)C3=O

InChI

1S/C20H18N4O2/c1-13-17(19(25)23(21-13)15-9-5-3-6-10-15)18-14(2)22-24(20(18)26)16-11-7-4-8-12-16/h3-12,17-18H,1-2H3

InChI key

FORCWSNQDMPPOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service