Skip to Content
Merck
All Photos(4)

Key Documents

T3260

Sigma-Aldrich

Toluidine Blue O

Powder

Synonym(s):

Basic Blue 17, Blutene chloride, Methylene Blue T50 or T extra, Tolonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN3S
CAS Number:
Molecular Weight:
305.83
Colour Index Number:
52040
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Toluidine Blue O, Technical grade

grade

technical grade

Quality Level

form

powder

composition

Dye content, ~80%

technique(s)

microbe id | staining: suitable

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Cc1cc2N=C3C=CC(\C=C3Sc2cc1N)=[N+](\C)C

InChI

1S/C15H15N3S.ClH/c1-9-6-13-15(8-11(9)16)19-14-7-10(18(2)3)4-5-12(14)17-13;/h4-8,16H,1-3H3;1H

InChI key

GEDVVYWLPUPJJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A metachromatic dye suitable for a wide variety of histological staining procedures.
Toluidine Blue O (TBO) is a thiazine dye of the quinone-imine family and is cationic in nature. It has been employed in polychromatic staining of paraffin embedded plant cell walls. It is a metachromatic dye suitable for a wide variety of histological staining procedures.

Application

Toluidine Blue O has been used to stain a variety of specimens like mesenchymal cells, taste buds, osteoblasts, matrix, etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arecoline Alters Taste Bud Cell Morphology, Reduces Body Weight, and Induces Behavioral Preference Changes in Gustatory Discrimination in C57BL/6 Mice
Wei-Hau Peng
Chemical Senses (2016)
Kyungho Yoon et al.
Ultrasound in medicine & biology, 45(9), 2391-2404 (2019-06-21)
Transcranial application of focused ultrasound (FUS) combined with vascular introduction of microbubble contrast agents (MBs) has emerged as a non-invasive technique that can temporarily create a localized opening in the blood-brain barrier (BBB). Under image-guidance, we administered FUS to sheep
Teaching Plant Anatomy Through Creative Laboratory Exercises null
Mrinalini Sharma et al.
Antimicrobial agents and chemotherapy, 52(1), 299-305 (2007-10-31)
Staphylococci are important causes of nosocomial and medical-device-related infections. Their virulence is attributed to the elaboration of biofilms that protect the organisms from immune system clearance and to increased resistance to phagocytosis and antibiotics. Photodynamic treatment (PDT) has been proposed
Polychromatic staining of plant cell walls by toluidine blue O
T.P.O'Brien
Protoplasma, 59(2) (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service