Merck
Copper-catalyzed C-H azidation of anilines under mild conditions.
Journal of the American Chemical Society (2012-11-08)
Conghui Tang, Ning Jiao
ABSTRACT
A novel and efficient copper-catalyzed azidation reaction of anilines via C-H activation has been developed. This method, in which the primary amine acts as a directing group by coordinating to the metal center, provides ortho azidation products regioselectively under mild conditions. This effective route for the synthesis of aryl azides is of great significance in view of the versatile reactivity of the azide products.
Product Description
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%