Skip to Content
Merck
All Photos(1)

Key Documents

01-3360

Sigma-Aldrich

4-Aminoantipyrine

≥99.0%, JIS special grade

Synonym(s):

4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O
CAS Number:
Molecular Weight:
203.24
Beilstein:
181635
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

availability

available only in Japan

mp

105-110 °C (lit.)

SMILES string

CN1N(c2ccccc2)C(=O)C(N)=C1C

InChI

1S/C11H13N3O/c1-8-10(12)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,12H2,1-2H3

InChI key

RLFWWDJHLFCNIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Teng et al.
Journal of hazardous materials, 190(1-3), 574-581 (2011-04-19)
4-Aminoantipyrine (AAP) is widely used in the pharmaceutical industry, in biochemical experiments and in environmental monitoring. AAP as an aromatic pollutant in the environment poses a great threat to human health. To evaluate the toxicity of AAP at the protein
A Z El-Sonbati et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 104, 213-221 (2012-12-27)
A novel series of copper(II) and palladium(II) with 4-derivatives benzaldehyde pyrazolone (L(n)) were synthesized. The mixed ligand complexes were prepared by using 1,10-phenanthroline (Phen) as second ligand. The structure of these complexes was identified and confirm by elemental analysis, molar
N Raman et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 85(1), 223-234 (2011-10-25)
Few novel 4-aminoantipyrine derived Schiff bases and their metal complexes were synthesized and characterized. Their structural features and other properties were deduced from the elemental analysis, magnetic susceptibility and molar conductivity as well as from mass, IR, UV-vis, (1)H NMR
Isela Lavilla et al.
Talanta, 98, 197-202 (2012-09-04)
In this work, a new method based on the combination of dispersive liquid-liquid microextraction (DLLME) with microvolume spectrophotometry has been developed as a greener and miniaturized alternative to the 5530 APHA standard method for determining phenols in water and wastewater.
Qihui Wang et al.
Journal of hazardous materials, 186(2-3), 1076-1081 (2010-12-21)
In this work, the immobilization of 4-aminoantipyrine onto bentonite was carried out and it was then used to investigate the adsorption behavior of Cr(III), Hg(II) and Pb(II) ions from aqueous solutions. The separation and preconcentration conditions of analytes were investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service