Skip to Content
Merck
All Photos(1)

Key Documents

06800

Supelco

4-Aminoantipyrine

for spectrophotometric det. of H2O2 and phenols, ≥98.0%

Synonym(s):

4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O
CAS Number:
Molecular Weight:
203.24
Beilstein:
181635
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (NT)
≥98.0%

quality

for spectrophotometric det. of H2O2 and phenols

technique(s)

UV/Vis spectroscopy: suitable
photometry: suitable

mp

105-110 °C (lit.)
108-110 °C

solubility

H2O: 0.1 g/mL, clear to slightly hazy

SMILES string

CN1N(c2ccccc2)C(=O)C(N)=C1C

InChI

1S/C11H13N3O/c1-8-10(12)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,12H2,1-2H3

InChI key

RLFWWDJHLFCNIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Cu-chelated polydopamine nanozymes with laccase-like activity for photothermal catalytic degradation of dyes.: This research highlights the synthesis of copper-chelated polydopamine nanozymes demonstrating laccase-like activities. These nanozymes enhance the photothermal degradation of dyes, providing a novel approach for environmental remediation (Wang et al., 2024).

  • Tramadol and M1 Bioavailability Induced by Metamizole Co-Administration in Donkeys (Equus asinus).: This study explores the pharmacokinetic interactions between tramadol and metamizole in donkeys, offering insights into veterinary medicine and drug administration practices (Araújo-Silva et al., 2024).

  • Pharmacokinetics-Based Pediatric Dose Evaluation and Optimization Using Saliva - A Case Study.: This paper presents a methodological innovation using saliva samples to optimize pediatric dosing of medications, which could revolutionize therapeutic drug monitoring in children (Anliker-Ort et al., 2024).

  • Spectrophotometric determination of phenol impurity in phenoxyethanol and phenol index of drinking water and municipal wastewater effluent after salting-out assisted liquid phase microextraction (SA-LPME).: This study offers a spectrophotometric method for detecting phenol impurities, enhancing water quality monitoring techniques (Roustaei et al., 2024).

  • A novel hybrid sensor array based on the polyphenol oxidase and its nanozymes combined with the machine learning based dual output model to identify tea polyphenols and Chinese teas.: This research introduces a hybrid sensor array employing polyphenol oxidase and its nanozymes for detecting tea polyphenols, showcasing the integration of enzymatic methods with machine learning for enhanced specificity and sensitivity in food analysis (Yang et al., 2024).

Other Notes

4-Aminoantipyrine has been used as a corrosion inhibitor of mild steel in 2 M HCl solution using different techniques: electrochemical impedance spectroscopy (EIS), potentiodynamic polarization and weight loss.[1]
Reagent for the determination of H2O2[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Saito et al.
Talanta, 34(7), 667-669 (1987-07-01)
Amberlite IRA 900 anion-exchange resin modified with manganese-tetrakis(sulphophenyl)-porphine has been used as a catalyst instead of peroxidase for the determination of hydrogen peroxide by the reaction 2H(2)O(2) + N,N-diethylaniline + 4-aminoantipyrine (catalyst)--> quinonoid dye (lambda(max) 550 nm) + 4H(2)O. The
Biphasic Variation Over Time in Presenting Features of Patients With COVID-19.
Nazreen Jamal et al.
Pediatrics, 146(5) (2020-09-05)
Alireza Radmanesh et al.
Radiology, 297(1), E223-E227 (2020-05-22)
Diffuse leukoencephalopathy and juxtacortical and/or callosal microhemorrhages were brain imaging features in critically ill patients with coronavirus disease 2019. Coronavirus disease 2019 (COVID-19) has been reported in association with a variety of brain imaging findings such as ischemic infarct, hemorrhage
Amy V Dora et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 73(3), 545-548 (2020-08-29)
We characterized serology following a nursing home outbreak of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) where residents were serially tested by reverse-transcription polymerase chain reaction (RT-PCR) and positive residents were cohorted. When tested 46-76 days later, 24 of 26
Stephanie Hackett et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 73(7), e2217-e2225 (2020-08-01)
Children living with human immunodeficiency virus (HIV) (CLHIV) receiving antiretroviral therapy (ART) in resource-limited settings are susceptible to high rates of acquired HIV drug resistance (HIVDR), but few studies include children initiating age-appropriate World Health Organization (WHO)-recommended first-line regimens. We

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service