Skip to Content
Merck
All Photos(1)

Key Documents

01-2180

Sigma-Aldrich

Alizarin Red S

SAJ special grade

Synonym(s):

3,4-Dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt, Alizarin Carmine, Alizarin Red, water soluble, Alizarin sodium monosulfonate, Alizarin sulfonate sodium, Alizarinsulfonic acid sodium salt, Sodium alizarinsulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7NaO7S
CAS Number:
Molecular Weight:
342.26
Colour Index Number:
58005
Beilstein:
4117090
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

grade

SAJ special grade

availability

available only in Japan

SMILES string

[Na+].Oc1c(O)c(cc2C(=O)c3ccccc3C(=O)c12)S([O-])(=O)=O

InChI

1S/C14H8O7S.Na/c15-11-6-3-1-2-4-7(6)12(16)10-8(11)5-9(22(19,20)21)13(17)14(10)18;/h1-5,17-18H,(H,19,20,21);/q;+1/p-1

InChI key

HFVAFDPGUJEFBQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S An et al.
Journal of periodontal research, 47(3), 374-382 (2011-12-06)
A number of bone-filling materials containing calcium (Ca(2+) ) and phosphate (P) ions have been used in the repair of periodontal bone defects; however, the effects that local release of Ca(2+) and P ions has on biological reactions are not
Xu Wang et al.
Electrophoresis, 34(2), 235-243 (2012-11-20)
A novel fluorescence detection system using a chemosensor for phosphoprotein in gel electrophoretic analysis has been developed. The system employed the alizarin red S-aluminum (III)-appended complex as a fluorescent staining dye to perform the convenient and selective detection of phosphorylated
Marjorie Zanini et al.
Journal of endodontics, 38(9), 1220-1226 (2012-08-16)
Biodentine (Septodont, Saint Maur des Faussés, France), a new tricalcium silicate-based cement, has recently been commercialized and advertised as a bioactive material. Its clinical application and physical properties have been widely described, but, so far, its bioactivity and biological effect
K N Speight et al.
Veterinary pathology, 50(2), 299-307 (2012-08-03)
The wild and captive koala population of the Mt Lofty Ranges in South Australia has a high level of renal dysfunction in which crystals consistent with calcium oxalate have been observed in the kidneys. This study aimed to describe the
Vishal Jhanji et al.
Current eye research, 37(2), 155-158 (2012-01-19)
To compare the quality of corneal endothelium of precut Descemet-stripping automated endothelial keratoplasty (DSAEK) tissue when transported with and without the anterior lamellar corneal tissue (ALCT) when organ-culture corneal storage methods are used. After microkeratome-assisted excision of anterior corneal lamella

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service