Skip to Content
Merck
All Photos(1)

Key Documents

11922416001

Roche

Anti-His6

from mouse IgG1

Synonym(s):

metal chelate affinity chromatography

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

form

lyophilized

packaging

pkg of 100 μg

manufacturer/tradename

Roche

isotype

IgG1

storage temp.

2-8°C

Related Categories

General description

Anti-His6 is a monoclonal antibody to His6-tagged proteins. Anti-His6 specifically recognizes an epitope of six consecutive histidine residues of both natural and recombinant proteins. The antibody reacts with native and denatured histidine-tagged fusion proteins independent of the location of the histidine-tag in the protein sequence.

Specificity

Anti-His6 specifically recognizes an epitope of six consecutive histidine residues (His6) in natural and recombinant proteins. It reacts with both native and denatured histidine-tagged fusion proteins, independent of location of the histidine-tag in the protein sequence. In addition, Anti-His6 preferentially recognizes the C-terminal His6-epitope with high sensitivity. Anti-His6 allows specific and sensitive detection of histidine-tagged proteins irrespective of the expression system used.
The antibody recognizes an epitope of six consecutive histidine residues (His6) in natural and recombinant proteins. It reacts with both native and denatured histidine-tagged fusion proteins, independent of the epitope-sequence location.

Application

Anti-His6 is used for the detection of His6-tagged proteins in:
  • Immunoblotting (e.g., dot blots and western blots)[1][2]
  • Immunoprecipitation
  • Immunoassays (ELISA)
  • Immunocytochemistry
  • Immunofluorescence

Preparation Note

Stabilizers: BSA
Working concentration: Working concentration of conjugate depends on application and substrate.
The following concentrations should be taken as a guideline:
  • ELISA: for detection 0.1 μg/ml; for coating 1 to 5 μg/ml
  • Immunoprecipitation: 0.5 to 5 μg/ml
  • Western blot: 0.2 to 0.5 μg/ml

Reconstitution

Add 1 ml double-distilled water to a final concentration of 100 μg/ml.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvatore Antonio Maria Cubisino et al.
Frontiers in molecular biosciences, 11, 1334819-1334819 (2024-04-12)
COVID-19, the infectious disease caused by the most recently discovered coronavirus SARS- CoV-2, has caused millions of sick people and thousands of deaths all over the world. The viral positive-sense single-stranded RNA encodes 31 proteins among which the spike (S)
Michael Kisiela et al.
The FEBS journal, 285(2), 275-293 (2017-11-21)
The human dehydrogenase/reductase SDR family member 4 (DHRS4) is a tetrameric protein that is involved in the metabolism of several aromatic carbonyl compounds, steroids, and bile acids. The only invertebrate DHRS4 that has been characterized to date is that from
Aniek D van der Woude et al.
Microbial cell factories, 15, 60-60 (2016-04-10)
Erythritol is a polyol that is used in the food and beverage industry. Due to its non-caloric and non-cariogenic properties, the popularity of this sweetener is increasing. Large scale production of erythritol is currently based on conversion of glucose by
Ji-Young Lim et al.
Cancer immunology research, 9(5), 503-513 (2021-03-28)
Progression of multiple myeloma is regulated by factors intrinsic to the clonal plasma cells (PC) and by the immune effector cells in the tumor microenvironment. In this study, we investigated the interaction between CD304 expression on myeloid-derived suppressor cells (MDSC)
Darija Putar et al.
Open biology, 14(1), 230372-230372 (2024-01-24)
RasG is a major regulator of macropinocytosis in Dictyostelium discoideum. Its activity is under the control of an IQGAP-related protein, IqgC, which acts as a RasG-specific GAP (GTPase activating protein). IqgC colocalizes with the active Ras at the macropinosome membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service