Skip to Content
Merck
All Photos(1)

Key Documents

11138600001

Roche

Interleukin-6, human (hIL-6)

recombinant (E. coli)

Synonym(s):

hIL-6, interleukin-6 human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>98% (SDS-PAGE)

form

solution

potency

<0.01 ng/mL EC50

mol wt

20,600 Da

packaging

pkg of 200,000 U (2 μg, 1 ml)

manufacturer/tradename

Roche

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | hybridoma: suitable

impurities

<0.1 EU/μgtested (LAL test)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL6(3569)

General description

Interleukin-6, human (hIL-6), is produced in E. coli and purified by standard chromatographic techniques.

Specificity

Recombinant IL-6, human is effective on mouse and human cells.
IL-6, a 26 kD protein, also known as B-cell stimulatory factor 2 (BSF-2, BCDF), interferon-β2 (IFN-β2), hybridoma/plasmacytoma growth factor (H(P)GF), and hepatocyte-stimulating factor is produced by many cell types (e.g., T-cells, monocytes, fibroblasts, endothelial cells). IL-6 induces the terminal maturation of activated B-cells into antibody-producing cells (B-cell differentiation activity. The hepatic acute phase response is induced by IL-6 (hepatocyte-stimulating activity). It was shown that IL-6 and IL-3 act synergistically to support the proliferation of hematopoietic stem cells (hemapoietic activity), however, IL-6 does not have antiviral activity, although it has been called IFN-β2. High affinity receptors for IL-6 are found on many cell types.

Application

Recombinant IL-6, human, is a growth factor for murine and human B-cells. It can be used to replace feeder cells in the preparation of murine and human hybridomas.

Biochem/physiol Actions

Interleukin-6 participates in several pleiotropic functions in various types of cells through its specific receptor (IL-6-R). It has been suggested that interleukin-6 may participate in the interaction between neuroendocrine and immune system by activating adrenocorticotropic hormone secretion. It has also been reported that IL-6 stimulates the bonding of receptor (IL-6-R) with a non-ligand-binding membrane glycoprotein, gp130.

Quality

Purity: 95% pure as determined by HPLC or SDS-PAGE [Endotoxin level: <0.1 EU/μg (LAL-test), <10 EU/ml (LAL-test).
Note: 1 EU corresponds to 0.1 ng

Sequence

Chain Length 184 AA
The primary structure of recombinant human IL-6 is identical to that of natural human IL-6 (one polypeptide chain, 184 amino acids), however, it contains an altered sequence within the first 15 amino-terminal amino acids.

Unit Definition

EC50 definition: The amount of hIL-6 that is required to support half-maximal stimulation of cell proliferation (MTT cleavage) with 7TD1 cells (1 unit equals <0.01 ng).

Physical form

Solution, filtered through 0.2 μm pore size membrane.

Preparation Note

Working concentration: 10 to 100 U/ml (0.1 1 ng/ml)
10 to 100 U/ml (0.1 to 1 ng/ml)
A concentration of 100 U/ml (1 ng/ml) is recommended for the preparation of B-cell hybridomas
Working solution: Dilute the concentrated IL-6 solution (200,000 U/ml) with PBS or culture medium containing 1 mg/ml BSA [or HSA (human serum albumin)] or 1 to 10% serum.
Storage conditions (working solution): -15 to -25 °C
Store the solution in aliquots at -15 to -25 °C.
Note: Avoid repeated freezing and thawing.

Reconstitution

Dilute the concentrated IL-6 solution (200,000 U/ml) with PBS or culture medium containing 1 mg/ml BSA [or HSA (human serum albumin)] or 1 to 10% serum.

Analysis Note

Specific activity/EC50: >1 x 108 U/mg, <0.01 ng/ml (hIL-6, NIBSC, interim standard, 88/514), at least the same specific activity (EC50) compared to the indicated standard is guaranteed.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Andus et al.
FEBS letters, 221(1), 18-22 (1987-08-31)
Conditioned medium from human monocytes contains a partially characterized hepatocyte-stimulating factor that simultaneously elevates the mRNA levels of the acute-phase protein beta-fibrinogen and decreases albumin mRNA in rat hepatoma cells. We demonstrate that recombinant human B-cell stimulatory factor 2, which
Human beta 2 interferon and B-cell differentiation factor BSF-2 are identical.
P B Sehgal et al.
Science (New York, N.Y.), 235(4790), 731-732 (1987-02-13)
A Muraguchi et al.
The Journal of experimental medicine, 167(2), 332-344 (1988-02-01)
The role of recombinant B cell stimulatory factor 2 (BSF-2/IL-6) in the regulation of growth and differentiation of B cells was investigated. rBSF-2 at 200 pg/ml could induce 50% of the maximum Ig production in B lymphoblastoid cell lines, the
Multiple actions of interleukin 6 within a cytokine network.
G G Wong et al.
Immunology today, 9(5), 137-139 (1988-05-01)
Interferon β2 as a promoter of growth and differentiation of B cells.
A Billiau
Immunology today, 8(3), 84-87 (1987-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service