Skip to Content
Merck
All Photos(1)

Key Documents

I2644

Sigma-Aldrich

IL-2 human

≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IL-2, Interleukin-2, Interleukin-2 human, T-cell growth factor, hIL-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

form

lyophilized powder

potency

≤700 EC50

quality

endotoxin tested

mol wt

15.5 kDa

packaging

pkg of 5X10 μg
pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL2(3558)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukin-2 (IL-2) is an α-helical cytokine belonging to T-helper cell type 1. It has a molecular weight of 15kDa. The IL2 gene is mapped to human chromosome 4q27. IL-2 is originally called as T cell growth factor.

Application

Interleukin-2 human has been used:
  • in the stimulation of natural killer cells
  • to study its ability to stimulate lymphocyte growth and prevent apoptosis
  • as an antigen in enzyme-linked immunosorbent assay (ELISA), to analyse the human autoantibody reactivities in health and in type 1 diabetes mellitus

Biochem/physiol Actions

Interleukin-2 (IL-2) is an immunomodulatory factor produced by certain subsets of T lymphocytes. This lympho­kine promotes long term growth of acti­vated T cells and related cell types. IL-2 plays a role in the activation and proliferation of NK cells, induces γ-inter­feron and B cell growth factor secretion, and modulates the expression of the IL-2 receptor.
Interleukin-2 (IL-2) is essential for proliferation of natural killer (NK) cells, lymphocytes and macrophages. It plays a key role in the immune system functions such as NK cell-stimulated immune responses. IL-2 also mediates the essential functions of regulatory T cells. It is implicated in autoimmune disease. IL-2 is associated with apoptosis and thus, possesses antitumor response. It mediates CD41+ T-cells differentiation into T-helper cell 1 and 2 effector subsets, preventing T-helper 17 differentiation.

Physical form

Lyophilized, 0.2 μm filtered, no additives.

Analysis Note

The biological activity is measured by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natural killer cells in the synovial fluid of rheumatoid arthritis patients exhibit a CD56bright, CD94bright, CD158negative phenotype
Pridgeon C, et al.
Rheumatology (Basel), 42(7), 870-878 (2003)
Stefania Braidotti et al.
Frontiers in bioscience (Landmark edition), 27(2), 39-39 (2022-03-01)
The T-cell engager antibody blinatumomab (BlincytoT⁢M) represents a promising rescue therapy for relapsed/refractory CD19+ acute lymphoblastic leukemia (B-ALL), although ~20-30% of patients still do not respond to treatment. Blinatumomab creates a tight synapsis between CD3+ T-lymphocytes and leukemic CD19+ B-cells
Loss of immune tolerance to IL-2 in type 1 diabetes
Perol L, et al.
Nature Communications, 7(2), 13027-13027 (2016)
Interleukin 2, 37-37 (2012)
Association of- 330 interleukin-2 gene polymorphism with oral cancer
Singh PK, et al.
The Indian Journal of Medical Research, 146(6), 730-730 (2017)

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service