Skip to Content
Merck
All Photos(1)

Key Documents

T3038

Sigma-Aldrich

Trizma® hydrochloride solution

1 M, BioReagent, for molecular biology

Synonym(s):

Tris hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

for molecular biology
for molecular biology

Quality Level

sterility

0.2 μm filtered

product line

BioReagent

form

solution

concentration

1 M

impurities

DNase, RNase, NICKase and protease, none detected

pH

8.0

application(s)

agriculture

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trizma® hydrochloride solution is a useful biological buffer.

Application

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per degree C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.
Trizma® hydrochloride solution may be used in the following studies:
  • As buffer for the 2-D electrophoresis of rat fibroblast cell.[1]
  • As buffer for the rapid isolation of high molecular weight plant DNA (50,000 base pairs or more in length).[2]
  • Selective immunoprecipitation of biotin-labeled DNA with antibiotin IgG and Staphylococcus sp.[3]

Other Notes

Prepared with pH-adjusted Biotechnology Performance Certified Trizma Base in 18 megohm water and 0.2 μm filtered.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Murray et al.
Nucleic acids research, 8(19), 4321-4325 (1980-10-10)
A method is presented for the rapid isolation of high molecular weight plant DNA (50,000 base pairs or more in length) which is free of contaminants which interfere with complete digestion by restriction endonucleases. The procedure yields total cellular DNA
P R Langer et al.
Proceedings of the National Academy of Sciences of the United States of America, 78(11), 6633-6637 (1981-11-01)
Analogs of dUTP and UTP that contain a biotin molecule covalently bound to the C-5 position of the pyrimidine ring through an allylamine linker arm have been synthesized. These biotin-labeled nucleotides are efficient substrates for a variety of DNA and
Emilie M F Kallenbach et al.
The Science of the total environment, 786, 147455-147455 (2021-05-09)
Chitinaceous organisms have been found to ingest microplastic; however, a standardised, validated, and time- and cost-efficient method for dissolving these organisms without affecting microplastic particles is still required. This study tested four protocols for dissolving organisms with a chitin exoskeleton:
Martin Dahl et al.
Environmental pollution (Barking, Essex : 1987), 273, 116451-116451 (2021-01-25)
Plastic pollution is emerging as a potential threat to the marine environment. In the current study, we selected seagrass meadows, known to efficiently trap organic and inorganic particles, to investigate the concentrations and dynamics of microplastics in their soil. We
Lisa W von Friesen et al.
Marine pollution bulletin, 142, 129-134 (2019-06-25)
Standardized methods for the digestion of biota for microplastic analysis are currently lacking. Chemical methods can be effective, but can also cause damage to some polymers. Enzymatic methods are known to be gentler, but often laborious, expensive and time consuming.

Related Content

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Questions

  1. What is the tris molarity in Trizma hydrochloride buffer cat# 41573 ?

    1 answer
    1. Trizma is a registered trademark. The concentration of this product is between 0.95 - 1.05 M. The exact concentration is listed on the lot specific Certificate of Analysis: https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/T3038-1LSLCQ0820.pdf.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service