SCT280
BioTracker™ Far Red Live Cell Cycle Assay Solution
Human
Synonym(s):
cell cycle dye, cell cycle indicator, cell cycle probe, live cell probe
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
NACRES:
NA.47
Product Name
BioTracker™ Far Red Live Cell Cycle Assay Solution,
biological source
human
Quality Level
packaging
vial of 50 tests
manufacturer/tradename
Millipore
technique(s)
cell culture | stem cell: suitable
flow cytometry: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Propidium iodide (PI) is often used for cell cycle analysis by flow cytometry. Compared to this analysis method, the BioTracker™ Far Red Live Cell Cycle Assay Solution does not require cell fixation or permeabilization, and is permeable in live cells. Use 5 uL of probe solution in 500 uL PBS for each sample. Vial contains enough for 50 tests.The far red probe uses the 633 laser, enabling multiplexing with shorter wavelength probes by freeing the 488 laser for probes that emit in the green range of the spectrum. IMPORTANT APPLICATION NOTES:*Wash cells thoroughly to remove trypsin. Residual trypsin can confound analysis of flow cytometry populations.*Adjust gates after cells are stained with probe SPECTRAL PROPERTIESExcitation: 633 - 647 nmEmission: 695 LP or 780/60 BP.A filter set for APC-Cy7 is suitable.
Features and Benefits
Far red fluorescence probe for analysis of cell cycle by flow cytometry.
Target description
The control system of the cell cycle is closely linked to cell proliferation. Cell division in which one cell produces two daughter cells occurs as part of the cell cycle. The cell cycle can be assessed by analyzing cells stained for DNA using flow cytometry, followed by analysis of the proportion of stained nuclei in cell cycle phase. The cell cycle is classified into two major phases: interphase and the mitotic (M) phase. Interphase is composed of G1 phase (preparation for DNA synthesis which occurs prior to cell division), S phase (DNA synthesis and replication), and G2 phase (a single cell with duplicated chromosomes). M phase involves mitosis and cytokinesis. Cell cycle progression is regulated by cyclins and cyclin-dependent kinases. To determine the action of anticancer drugs and others, it is important to analyze the mechanism of cell-cycle checkpoint.
Storage and Stability
Store BioTracker™ Far Red Live Cell Cycle Assay Solution at -20 °C, protected from light.
Other Notes
Live cell probe for fluorescent cell cycle analysis. Unlike propidium iodide, this probe is membrane permeable and highly specific for DNA, which makes it possible to analyze the cell cycle by simply adding the reagent to a cell suspension.
Legal Information
BioTracker is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service