S7903-M
Amplifluor® UniPrimer II Fluorescein
Synonym(s):
Amplifluor Primer, Fluorescein UniPrimer, UniPrimer II Fluorescein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.21
Recommended Products
manufacturer/tradename
Chemicon®
Quality Level
100
300
technique(s)
PCR: suitable
application(s)
genomic analysis
detection method
fluorometric
Related Categories
General description
Fluorescently labeled primers are used as probes to detect the amplified PCR product. It is used to produce the amplicon. These probes are designed such that the primer extension leads to improved fluorescence.
The Amplifluor® UniPrimer-fluorescein is a hairpin primer labeled with a fluorescent energy transfer pair (fluorescein and non-fluorescent quencher) and a 3′ single-stranded oligonucleotide tail and is provided at a concentration of 5.0 μM (25.0 mL).
Fluorescence (Linear:Hairpin): 39. Excitation = 495nm; Emission = 516nm.
Fluorescence (Linear:Hairpin): 39. Excitation = 495nm; Emission = 516nm.
Physical form
5.0μM (25.0mL)
Legal Information
Amplifluor is a registered trademark of Merck KGaA, Darmstadt, Germany
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rohan T Ranasinghe et al.
Chemical communications (Cambridge, England), (44), 5487-5502 (2005-12-17)
Synthetic chemistry has been central to the design of modern methods of genetic analysis. In this article, we discuss the underlying chemistry and biophysical principles that have been used in the development of robust methods for the analysis of DNA
Wim Reybroeck et al.
Journal of agricultural and food chemistry, 58(12), 7258-7265 (2010-05-27)
A liquid chromatographic tandem mass spectrometric method for the determination of sulfa drugs in beeswax was developed. When performing residue control on beeswax intended for the fabrication of wax foundations, residues of sulfonamides were found. A migration test was set
Séverine Remy et al.
Methods in molecular biology (Clifton, N.J.), 597, 109-125 (2009-12-17)
Lentiviral vectors are now well recognized as good vehicles for gene delivery. This is because they can efficiently transduce both dividing and post-mitotic cells, and stably integrate into the host genome allowing for long-term expression of the transgene. Their potential
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service