Skip to Content
Merck
All Photos(1)

Key Documents

S7150

Sigma-Aldrich

OxyBlot Protein Oxidation Detection Kit

The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.

Synonym(s):

OxyBlot assay, Protein oxidation kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

manufacturer/tradename

Chemicon®
OxyBlot

technique(s)

western blot: suitable

detection method

chemiluminescent

shipped in

wet ice

General description

Oxidative modification of proteins by oxygen free radicals and other reactive species occurs in physiologic and pathologic processes. As a consequence of the modification, carbonyl groups are introduced into protein side chains by a site-specific mechanism. The OxyBlot Kit provides reagents for simple and sensitive immunodetection of these carbonyl groups, which is a hallmark of the oxidation status of proteins.

Application

Preparation of Cell Lysate

1. Wash adherent cells twice in the dish or flask with ice-cold PBS and drain off PBS. Wash non-adherent cells in PBS and centrifuge at 800 to 1000 rpm in a table-top centrifuge for 5 minutes to pellet the cells.
2. Add ice-cold RIPA buffer to cells (1 ml per 107 cells/100 mm dish/150 cm2 flask; 0.5 ml per 5 x 106 cells/60 mm dish/75 cm2 flask).
3. Scrape adherent cells off the dish or flask with either a rubber policeman or a plastic cell scraper that has been cooled in ice-cold distilled water. Transfer the cell suspension into a centrifuge tube. Gently rock the suspension on either a rocker or an orbital shaker in the cold room for 15 minutes to lyse cells.
4. Centrifuge the lysate at 14,000 x g in a precooled centrifuge for 15 minutes. Immediately transfer the supernatant to a fresh centrifuge tube and discard the pellet.
5. Dilute the cell lysate at least 1 : 10 before determining the protein concentration because of the interference of the detergents in the lysis buffer with the Coomassie-based reagent. At this step, the sample can be divided into aliquots and stored at -20¡C for up to a month.
TIP: When working with large volumes of non-adherent cells, the cells may not be cooled quickly enough to maintain the activity of the protein being studied. In this case, pour the cell suspension into a mixture of an equal mass of 2 x PBS and ice, then collect the cells by centrifugation and perform the lysis as described above.

RIPA lysis buffer:
Catalog number 20-188
http://www.millipore.com/catalogue/item/20-188

Reducing agent is very important for preventing oxidation:
The protein lysate may be prepared using a variety of protocols. Most standard buffers such as RIPA and Triton are also suitable. It is recommended that a reducing agent be added to the lysis buffer to prevent the oxidation of proteins that may occur after cell lysis. Lysis buffer containing either 1-2% 2-mercaptoethanol or 50 mM DTT sufficiently inhibits this oxidation, but has no adverse effect on the derivatization reaction in the OxyBlot protocol. Purified protein samples are also suitable for analysis with the OxyBlot Kit.

For general information about protein extraction, please check this link.

http://www.millipore.com/immunodetection/id3/proteinextraction

Storage and Stability

Store the protein standard (component 90450) at -25° to -15°C upon receipt. Store all other components at 2°C to 8°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ketone bodies stimulate chaperone-mediated autophagy.
Finn, Patrick F and Dice, J Fred
The Journal of Biological Chemistry, 280, 25864-25870 (2005)
Gamma-glutamylcysteine ethyl ester protection of proteins from Abeta(1-42)-mediated oxidative stress in neuronal cell culture: a proteomics approach
Boyd-Kimball, D. et al.
Journal of Neuroscience Research, 79(5), 707-713 (2005)
A transient antioxidant stress response accompanies the onset of disuse atrophy in human skeletal muscle.
Dalla Libera, Luciano, et al.
Journal of Applied Physiology, 107, 549-557 (2009)
David Bernhard et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 19(9), 1096-1107 (2005-06-30)
Smoking is a significant risk factor for development of atherosclerosis. However, the pathophysiology of smoking-mediated vessel wall damage is not understood. With tools ranging from analytical chemistry to cell biology, we show that cigarette smoke contains metals that catalyze the
Proteasome inhibition by MG-132 induces apoptotic cell death and mitochondrial dysfunction in cultured rat brain oligodendrocytes but not in astrocytes.
Olaf Goldbaum, Grit Vollmer, Christiane Richter-Landsberg
Glia null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service