Skip to Content
Merck
All Photos(2)

Key Documents

MABT401

Sigma-Aldrich

Anti-Lubricin Antibody/Proteoglycan 4, clone 9G3

clone 9G3, from mouse

Synonym(s):

Proteoglycan 4, Lubricin, Megakaryocyte-stimulating factor, Superficial zone proteoglycan, Proteoglycan 4 C-terminal part

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9G3, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRG4(10216)

General description

The protein called Lubricin/Proteoglycan 4 is an important secreted protein in the joint critical for joint function and thus is also a focus of research in joint diseases as well. Lubricin/Proteoglycan 4 plays a role in boundary lubrication and prevents the deposition of cartilage from the synovial fluid by controlling the synovial growth of the synovial cells and inhibiting the adhesion of the cells to the cartilage surface. During development, Lubricin/Proteoglycan 4 also acts as a growth factor for hematopoietic and endothelial cell lineages, and Lubricin/Proteoglycan 4 appears important for proper barrier and lubrication development in tissues like the eye. Lubricin/Proteoglycan 4 is highly expressed in joints and near joint boundaries but can also be found in the brain, heart, lung, kidney and other tissues. Lubricin/Proteoglycan 4 has several isoforms that differ in their cell and tissue expression. Isoform C and D are the most widely expressed forms. Finally because of its effects on surfaces, Lubricin/Proteoglycan 4 is being examined in tissue engineering and artificial cartilage development as a treatment for joint diseases.

Immunogen

Recombinant protein corresponding to human Lubricin/Proteoglycan 4.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Lubricin/Proteoglycan 4 in human liver and human heart tissue.
Immunohistochemistry Analysis: A representative lot detected Lubricin/Proteoglycan 4 in articular cartilage tissue (Elsaid, K.A, et al. (2012). Osteoarthritis and Cartilage. 20:940-948).
Immunohistochemistry Analysis: A representative lot detected Lubricin/Proteoglycan 4 in rat arterial cartilage tissue (Jay, G.D, et al. (2012). Arthritis & Rheumatism. 64(4):940-948).
Immunohistochemistry Analysis: A representative lot detected Lubricin/Proteoglycan 4 in articular cartilage from rat
(Manuscript in preparation).
Western Blotting Analysis: A representative lot detected Lubricin/Proteoglycan 4 in lubricin purified from synovial fluid of patients (Manuscript in preparation).
This Anti-Lubricin Antibody/Proteoglycan 4, clone 9G3 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of Lubricin/Proteoglycan 4.

Quality

Evaluated by Western Blotting in Lubricin purified protein.

Western Blotting Analysis: 0.02 µg/mL of this antibody detected Lubricin/Proteoglycan 4 in 1 µg of Lubricin purified protein.

Target description

~220 kDa observed. The calculated molecular weight of this protein is 151 kDa with many isoforms observed around 145 kDa and it can run higher due to glycosylation.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong Li et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 9(21), e2105909-e2105909 (2022-04-19)
Diseases of the knee joint such as osteoarthritis (OA) affect all joint elements. An in vitro human cell-derived microphysiological system capable of simulating intraarticular tissue crosstalk is desirable for studying etiologies/pathogenesis of joint diseases and testing potential therapeutics. Herein, a
R M Irwin et al.
Osteoarthritis and cartilage, 28(4), 492-501 (2020-02-28)
Viscosupplementation has been used for decades to treat mild to moderate osteoarthritis, yet it is unknown if the lubricating function of different pathological synovial fluids (SF) vary, or if they respond differentially to viscosupplementation. The objectives of this study were
Elizabeth Feeney et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 37(5), 1071-1079 (2019-03-13)
The objective of this study was to examine temporal variations in synovial fluid composition and lubrication following articular fracture. Post-traumatic osteoarthritis (PTOA) was induced by creating an osteochondral fracture in the middle carpal joint of four horses while the contralateral
Tryptase I? regulation of joint lubrication and inflammation via proteoglycan-4 in osteoarthritis.
Das, et al.
Nature Communications, 14, 1910-1910 (2023)
Amanda Watkins et al.
BMC veterinary research, 17(1), 189-189 (2021-05-14)
Lameness is a debilitating condition in equine athletes that leads to more performance limitation and loss of use than any other medical condition. There are a limited number of non-terminal experimental models that can be used to study early inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service