Skip to Content
Merck
All Photos(1)

Key Documents

MABT219

Sigma-Aldrich

Anti-Actin Antibody, clone JLA20

clone JLA20, from mouse

Synonym(s):

Actin, alpha skeletal muscle, Alpha-actin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

JLA20, monoclonal

species reactivity

human

species reactivity (predicted by homology)

chicken (immunogen homology)

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Actin is an abundantly expressed in eukaryotic cells and it is the core component of the cytoskeleton. It exists in three isoforms: alpha actin, which is found in muscle tissue and forms fibrous actin that facilitates muscle contractility; and beta and gamma actin which are found in non-muscle cells and facilitate cell motility. Actin exists initially as globular actin (G-actin) which is converted to filamentous actin (F-actin) upon assembly. The formation of F-actin is a complex multistep process that is tightly regulated by signaling from the actin-binding proteins (ABPs) and Rho family GTPases. Filamentous actin is crosslinked with other actin polymers to form a complex cytoskeletal network which is required for exo- and phagocytosis, vesicular trafficking, cell migration, and gene regulation, among other cellular processes.

Application

Detect Actin using this Anti-Actin Antibody, clone JLA20 validated for use in Western Blotting, Immunofluorescence.
Immunofluorescence Analysis: A representative lot was used by an independent laboratory in NRK cells. (Cao, L.G., et al. (1993). The Journal of Cell Biology. 123(1):173-181.)

Western Blot Analysis: A representative lot was used by an independent laboratory in Me4405 melanoma cell lysate. (Thomas, W.D., et al. (2000). Journal of Immunology. 165:5612–5620.)

Quality

Evaluated by Western Blot in SK-BR3 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Actin on 10 µg of SK-BR3 cell lysate.

Target description

~42 kDa observed

Linkage

Replaces: 04-1040

Physical form

Format: Purified

Analysis Note

Control
SK-BR3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lívia X S Farah et al.
American journal of physiology. Renal physiology, 310(2), F123-F127 (2015-10-09)
Glucagon like peptide-1 (GLP-1) is an incretin hormone constantly secreted from the intestine at low basal levels in the fasted state; plasma concentrations rise rapidly after nutrient ingestion. Upon release, GLP-1 exerts insulinotropic effects via a G protein-coupled receptor, stimulation
David J Hughes et al.
Journal of virology, 86(2), 1034-1045 (2011-11-11)
Establishment of persistent Epstein-Barr virus (EBV) infection requires transition from a program of full viral latency gene expression (latency III) to one that is highly restricted (latency I and 0) within memory B lymphocytes. It is well established that DNA
Tanja Spoettl et al.
World journal of gastroenterology, 13(7), 1032-1041 (2007-03-22)
To examine the factor(s) involved in differentiation of intestinal macrophages (IMACs) using a recently established in vitro model. To test whether soluble or membrane bound factors induce IMAC-differentiation, freshly elutriated monocytes (MO) were incubated with conditioned media or cell membranes
Chunhua Zhang et al.
Frontiers in plant science, 4, 238-238 (2013-07-23)
In plant cells the actin cytoskeleton adopts many configurations, but is best understood as an unstable, interconnected track that rearranges to define the patterns of long distance transport of organelles during growth. Actin filaments do not form spontaneously; instead filament
Jennifer Huggins et al.
Blood, 109(4), 1611-1619 (2006-10-13)
Unmethylated CpG DNA activation of naive CD27- B cells has been reported to require B-cell-receptor (BCR) cross-linking. We describe a culture system using CpG DNA with sequential steps for T-cell-independent activation of naive CD19+CD27- human peripheral blood B cells that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service