Skip to Content
Merck
All Photos(2)

Key Documents

MAB2624

Sigma-Aldrich

Anti-Endomucin Antibody, clone V.5C7

clone V.5C7, from rat

Synonym(s):

Endomucin-2, Gastric cancer antigen Ga34, Mucin-14, MUC-14, Endomucin-1/2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

V.5C7, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Emcn(59308)

General description

Endomucin is an endothelial sialomucin belonging to a class of glycoproteins characterized by their high levels of O-glycosolation and rich quantities of sialic acid. It inhibits focal adhesion assembly and hinders extracellular matrix and cell interaction. Observed expression is highest in kidney heart, brain, liver, lung, spleen, and thymus. Endomucin is only found in endothelial cells.

Immunogen

Mouse endothelial cells

Application

Anti-Endomucin Antibody, clone V.5C7 detects level of Endomucin & has been published & validated for use in WB, IP, IH, FC.
Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP of bEND cell lysate (Morgan, 1999).

Immunohistochemistry Analysis: A previous lot was used by an independent laboratory in immunhistochemical staining of frozen and paraffin-embedded tissue sections of mouse tongue, heart, uterus, brain, and small intestine (Morgan, 1999).

Flow Cytometry Analysis: A previous lot was used by an independent laboratory in FC of transfected COS-7 cells (Morgan, 1999).
Research Sub Category
Organelle & Cell Markers

Adhesion (CAMs)

Quality

Evaluated by Western Blot on mouse endothelioma cell line, bEND3, cell lysate.

Western Blot Analysis: 0.0025 µg/mL of this antibody detected Endomucin in 10 µg of bEND3 cell lysate.

Target description

~ 75kDa observed

Physical form

Format: Purified

Analysis Note

Control
bEND3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kate Crawford et al.
Cancer biology & therapy, 22(10-12), 619-629 (2021-12-10)
Standard chemotherapy regimens for gastric adenocarcinoma (GAC) have limited efficacy and considerable toxicity profiles. Nab-paclitaxel has shown promising antitumor benefits in previous GAC preclinical studies. Dovitinib inhibits members of the receptor tyrosine kinase family including FGFR, VEGFR and PDGFR, and
Enhancing gastric cancer conventional chemotherapy effects by triple angiokinase inhibitor nintedanib in preclinical models.
Awasthi, et al.
Frontiers in Oncology, 13, 1145999-1145999 (2023)
Meghan Grojean et al.
Journal of cellular and molecular medicine, 25(11), 4950-4961 (2021-05-04)
Elevated expression of multiple growth factors and receptors including c-Met and VEGFR has been reported in gastric adenocarcinoma (GAC) and thus provides a potentially useful therapeutic target. The therapeutic efficacy of foretinib, a c-Met/VEGFR2 inhibitor, was determined in combination with
Kristina Y Aguilera et al.
Bio-protocol, 4(19) (2014-10-05)
Therapy-induced hypoxia drives changes in the tumor microenvironment that contribute to the poor response to therapy. Hypoxia is capable of driving the expression and/or activation of specific signaling cascades (e.g., c-Met, Axl, CTGF), the recruitment of tumor promoting immune cells
Priya Balasubramanian et al.
Nutrients, 16(7) (2024-04-13)
High-fat diets (HFDs) have pervaded modern dietary habits, characterized by their excessive saturated fat content and low nutritional value. Epidemiological studies have compellingly linked HFD consumption to obesity and the development of type 2 diabetes mellitus. Moreover, the synergistic interplay

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service