Skip to Content
Merck
All Photos(2)

Key Documents

L2140

Sigma-Aldrich

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia)

Isolectin B4 (BSI-B4), biotin conjugate, lyophilized powder

Synonym(s):

Bandeirea simplicifolia agglutinin, BS-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

Bandieraea simplicifolia

Quality Level

conjugate

biotin conjugate

form

lyophilized powder

potency

<50 μg per mL agglutination activity (using human blood group B erythrocytes)

composition

Protein, ~95% E1%/280

extent of labeling

3-6 mol biotin per mol protein

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia) has A and B subunits, which recognizes αGalNAc and αGal-end groups respectively. Lectins are extracted from the seeds of the legume shrub Griffonia simplicifolia, which are carbohydrate binding proteins.

Application

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia) has been used for immunohistochemistry and immunofluorescence staining.

Biochem/physiol Actions

BS-I has a major affinity for terminal α-D-galactosyl residues with a secondary affinity for terminal N-acetyl-α-D-galactosaminyl residues.
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia) agglutinates human blood group A and B cells. It is specific for αGalNAc- and αGal. It acts as a reagent for the detection of this αGal epitope in biological materials. Lectin from Bandeiraea simplicifolia is used as a marker for xenoreactive antigen (αGal1−3Gal) and cancer. Lectin plays an important role in fertilization, embryogenesis, inflammation, metastasis and host-parasite recognition.

Other Notes

BS-I is a tetrameric lectin consisting of two types of subunits designated A and B. There are five BS-I isolectins with different subunit composition: BSI-B4, BSI-AB3, BSI-A2B2, BSI-A3B and BSI-A4. BSI-B4 is blood group B specific and has an exclusive affinity for terminal α-D-galactosyl residues, whereas BSI-A4 has blood group A specificity and has a major affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Physical form

Contains citrate buffer salts and calcium chloride

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution using phosphate buffered saline, pH 6.8, containing, for each lectin, calcium, magnesium, and manganese at different concentrations. This activity is the lowest concentration to agglutinate a 2% suspension of appropriate erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quan Yuan et al.
Tissue engineering. Part A, 24(9-10), 719-728 (2017-10-06)
The aim of this study was to analyze the three-dimensional distribution of hypoxia in the arteriovenous (AV) loop model in rats, by examining the distribution of hypoxia-inducible factor-1 alpha (HIF-1α). AV loops were created from the femoral artery and vein
Sensory processing of deep tissue nociception in the rat spinal cord and thalamic ventrobasal complex
Sikandar S, et al.
Physiological Reports, 5(14), e13323-e13323 (2017)
Ramin Raouf et al.
Science signaling, 11(535) (2018-06-21)
Mechanically activated, slowly adapting currents in sensory neurons have been linked to noxious mechanosensation. The conotoxin NMB-1 (noxious mechanosensation blocker-1) blocks such currents and inhibits mechanical pain. Using a biotinylated form of NMB-1 in mass spectrometry analysis, we identified 67
Peripheral delta opioid receptors mediate duloxetine antiallodynic effect in a mouse model of neuropathic pain
Ceredig RA, et al.
The European Journal of Neuroscience, 48(5), 2231-2246 (2018)
The Lectins: Properties, Functions, and Applications in Biology and Medicine (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service