MAB8186
Anti-EBV EA-D-p52/50 Antibody, clone R3
clone R3, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
R3, monoclonal
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
immunofluorescence: suitable
western blot: suitable
isotype
IgG1
shipped in
wet ice
Specificity
EA-D-p52\50
Application
Anti-EBV EA-D-p52/50 Antibody, clone R3 detects level of EBV EA-D-p52/50 & has been published & validated for use in ELISA, IF & WB.
Immunofluorescence
ELISA
Immunoblotting
Optimal dilutions must be determined by the end user.
ELISA
Immunoblotting
Optimal dilutions must be determined by the end user.
Research Category
Infectious Diseases
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Infectious Diseases - Viral
Physical form
Format: Purified
Purified immunoglobulin. Liquid in 0.02M PBS, 0.25M NaCl, 0.1% sodium azide.
Storage and Stability
Maintain at 2-8°C.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Functions of the Epstein-Barr virus EBNA1 protein in viral reactivation and lytic infection.
Sivachandran, N; Wang, X; Frappier, L
Journal of virology null
Epstein-Barr virus EBNA1 protein regulates viral latency through effects on let-7 microRNA and dicer.
Mansouri, S; Pan, Q; Blencowe, BJ; Claycomb, JM; Frappier, L
Journal of virology null
Identification of polypeptide components of the Epstein-Barr virus early antigen complex with monoclonal antibodies.
Pearson, G R, et al.
Journal of virology, 47, 193-201 (1983)
Epstein-Barr virus protein kinase BGLF4 is a virion tegument protein that dissociates from virions in a phosphorylation-dependent process and phosphorylates the viral immediate-early protein BZLF1.
Asai, R; Kato, A; Kato, K; Kanamori-Koyama, M; Sugimoto, K; Sairenji, T; Nishiyama, Y; Kawaguchi, Y
Journal of virology null
Association of Epstein-Barr virus early antigen diffuse component and virus-specified DNA polymerase activity.
Li, J S, et al.
Journal of virology, 61, 2947-2949 (1987)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service